Merck
  • Identification of pancreatic glycoprotein 2 as an endogenous immunomodulator of innate and adaptive immune responses.
Identification of pancreatic glycoprotein 2 as an endogenous immunomodulator of innate and adaptive immune responses.
Journal of immunology (Baltimore, Md. : 1950) (2012-08-15)
Lael Werner, Daniela Paclik, Christina Fritz, Dirk Reinhold, Dirk Roggenbuck, Andreas Sturm
ABSTRACT
Pancreatic autoantibodies are Crohn disease-specific serologic markers. The function and immunological role of their recently identified autoantigen, glycoprotein 2 (GP2), are unknown. We therefore investigated the impact of GP2 on modulation of innate and adaptive immune responses to evaluate its potential therapeutic use in mucosal inflammation. Our data indicate a previously unknown function for GP2 as an immunomodulator. GP2 was ubiquitously expressed on cells vital to mucosal immune responses. The expression of GP2 was upregulated on activated human T cells, and it was further influenced by pharmaceutical TNF-α inhibitors. Recombinant GP2 significantly decreased human intestinal epithelial cells, mucosal and peripheral T cell proliferation, apoptosis, and activation, and it distinctly modulated cytokine secretion. Furthermore, intestinal epithelial cells stimulated with GP2 potently attracted T cells. In conclusion, we demonstrate a novel role for GP2 in immune regulation that could provide a platform for new therapeutic interventions in the treatment of Crohn disease.
Sigma-Aldrich
Anti-GP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.