Merck
  • New pyridin-3-ylmethyl carbamodithioic esters activate pyruvate kinase M2 and potential anticancer lead compounds.
New pyridin-3-ylmethyl carbamodithioic esters activate pyruvate kinase M2 and potential anticancer lead compounds.
Bioorganic & medicinal chemistry (2015-06-18)
Yu Zhang, Bin Liu, Xingyu Wu, Ridong Li, Xianling Ning, Yu Liu, Zhenming Liu, Zemei Ge, Runtao Li, Yuxin Yin
ABSTRACT
Pyruvate kinase M2 (PKM2) is a key protein responsible for cancer's Warburg effect. Activation of PKM2 may alter aberrant metabolism in cancer cells, which suggests PKM2 as a tumor selective therapeutic target. In this paper, the lead compound 8 was first discovered as a new kind of PKM2 activator from a random screening of an in-house compound library. Then, a series of lead compound 8 analogs were designed, synthesized and evaluated for their activation of PKM2 and anticancer activities. 7-Azaindole analog 32 was identified as the most potent PKM2 activator. Compounds with potent enzyme activity also exhibited selective anti-proliferation activity on cancer cell lines HCT116, Hela and H1299 compared with non-tumor cell line BEAS-2B. The structure-activity relationships of these compounds were supported by molecular docking results. Preliminary pharmacological studies also showed that compound 32 arrests the cell cycle at the G2/M phase in HCT116 cell line.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Tetramethylsilane, ≥99.0% (GC)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Phospho(enol)pyruvic acid trisodium salt hydrate, ≥97% (enzymatic)
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis