Merck
  • Elevated miR-122 serum levels are an independent marker of liver injury in inflammatory diseases.
Elevated miR-122 serum levels are an independent marker of liver injury in inflammatory diseases.
Liver international : official journal of the International Association for the Study of the Liver (2014-07-22)
Christoph Roderburg, Fabian Benz, David Vargas Cardenas, Alexander Koch, Joern Janssen, Mihael Vucur, Jeremie Gautheron, Anne T Schneider, Christiane Koppe, Karina Kreggenwinkel, Henning W Zimmermann, Mark Luedde, Christian Trautwein, Frank Tacke, Tom Luedde
ABSTRACT
Serum concentrations of miR-122 were proposed as a marker for various inflammatory diseases, but the mechanisms driving alterations in miR-122 serum levels are unknown. We analysed miR-122 serum levels and hepatic miR-122 expression in mice after hepatic ischaemia and reperfusion (I/R) injury. These data were compared with data from mice after caecal pole ligation and puncture (CLP) procedure. To translate these data into the human, we analysed miR-122 serum concentrations in a cohort of 223 patients with critical illness and 57 patients with cirrhosis. We detected strongly elevated levels of miR-122 in mice after hepatic I/R injury. miR-122-concentrations correlated with the degree of liver damage according to AST/ALT and were associated with the presence of hepatic cell death detected by TUNEL staining. miR-122 levels were elevated in the cellular supernatants in an in vitro model of hepatocyte injury, supporting the hypothesis that the passive release of miR-122 represents a surrogate for hepatocyte death in liver injury. Moreover, miR-122 levels were almost normal in patients with cirrhosis without ongoing liver damage, but were elevated when liver injury was present. In contrast to previous assumptions, miR-122-concentrations were independent of the presence of infection/sepsis in mice or human patients. miR-122 levels did not correlate with disease severity or mortality in critically ill patients. In contrast, serum miR-122 levels strictly correlated with the presence of hepatic injury in these patients. In mice and humans, miR-122 levels represent an independent and potent marker of ongoing liver injury and hepatic cell death regardless of the underlying disease.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, ≥99%
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Fluorescein (free acid), Dye content 95 %
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Ethanol, purum, secunda spirit, denaturated with 2% 2-butanone, S15, ~96% (based on denaturant-free substance)
Supelco
2-Propanol, analytical standard
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Phenol solution, BioReagent, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, for molecular biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, for molecular biology, BioReagent, ≥99.5%
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material