Merck
  • Gram-negative prosthetic joint infection: outcome of a debridement, antibiotics and implant retention approach. A large multicentre study.
Gram-negative prosthetic joint infection: outcome of a debridement, antibiotics and implant retention approach. A large multicentre study.
Clinical microbiology and infection : the official publication of the European Society of Clinical Microbiology and Infectious Diseases (2014-04-29)
D Rodríguez-Pardo, C Pigrau, J Lora-Tamayo, A Soriano, M D del Toro, J Cobo, J Palomino, G Euba, M Riera, M Sánchez-Somolinos, N Benito, M Fernández-Sampedro, L Sorli, L Guio, J A Iribarren, J M Baraia-Etxaburu, A Ramos, A Bahamonde, X Flores-Sánchez, P S Corona, J Ariza
ABSTRACT
We aim to evaluate the epidemiology and outcome of gram-negative prosthetic joint infection (GN-PJI) treated with debridement, antibiotics and implant retention (DAIR), identify factors predictive of failure, and determine the impact of ciprofloxacin use on prognosis. We performed a retrospective, multicentre, observational study of GN-PJI diagnosed from 2003 through to 2010 in 16 Spanish hospitals. We define failure as persistence or reappearance of the inflammatory joint signs during follow-up, leading to unplanned surgery or repeat debridement>30 days from the index surgery related death, or suppressive antimicrobial therapy. Parameters predicting failure were analysed with a Cox regression model. A total of 242 patients (33% men; median age 76 years, interquartile range (IQR) 68-81) with 242 episodes of GN-PJI were studied. The implants included 150 (62%) hip, 85 (35%) knee, five (2%) shoulder and two (1%) elbow prostheses. There were 189 (78%) acute infections. Causative microorganisms were Enterobacteriaceae in 78%, Pseudomonas spp. in 20%, and other gram-negative bacilli in 2%. Overall, 19% of isolates were ciprofloxacin resistant. DAIR was used in 174 (72%) cases, with an overall success rate of 68%, which increased to 79% after a median of 25 months' follow-up in ciprofloxacin-susceptible GN-PJIs treated with ciprofloxacin. Ciprofloxacin treatment exhibited an independent protective effect (adjusted hazard ratio (aHR) 0.23; 95% CI, 0.13-0.40; p<0.001), whereas chronic renal impairment predicted failure (aHR, 2.56; 95% CI, 1.14-5.77; p 0.0232). Our results confirm a 79% success rate in ciprofloxacin-susceptible GN-PJI treated with debridement, ciprofloxacin and implant retention. New therapeutic strategies are needed for ciprofloxacin-resistant PJI.
Product Description
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Ciprofloxacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Ciprofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)