Merck
  • Hollow fiber based liquid-liquid-liquid microextraction combined with sweeping micellar electrokinetic chromatography for the sensitive determination of second-generation antidepressants in human fluids.
Hollow fiber based liquid-liquid-liquid microextraction combined with sweeping micellar electrokinetic chromatography for the sensitive determination of second-generation antidepressants in human fluids.
The Analyst (2015-01-27)
Xiaoqing Zhou, Man He, Beibei Chen, Bin Hu
ABSTRACT
An effective dual preconcentration method involving off-line hollow fiber liquid-liquid-liquid microextraction (HF-LLLME) and on-line sweeping micellar electrokinetic chromatography (sweeping-MEKC) was proposed for the determination of five second-generation antidepressants, including fluoxetine, sertraline, paroxetine, fluvoxamine and citalopram. In HF-LLLME, the analytes were extracted from the sample solution into phenetole impregnated in the pores of the hollow fiber and then back-extracted into 10 μL 0.1 mol L(-1) HAc inside the hollow fiber. Then, the acceptor phase was spiked with 2.8 μL isopropanol (IPA) and introduced into CE for sweeping. In the sweeping-MEKC process, five target analytes were separated in less than 15 min with a background electrolyte consisting of 76% (v/v) 50 mmol L(-1) citric acid (pH 2.2) containing 100 mmol L(-1) sodium dodecyl sulfate (SDS) and 24% (v/v) IPA. The hydrodynamic injection was performed at 50 mbar for 140 s. Under optimized conditions, the limits of detection were in the range of 0.40-1.55 μg L(-1) with enrichment factors of 1897- to 5952-fold for target analytes, with a dynamic linear range of 0.6/5.0-200 μg L(-1). The developed method demonstrated excellent clean-up ability and high enrichment factors and was successfully applied to the analysis of target analytes in human urine and plasma samples.
Sigma-Aldrich
1-Octanol, natural, ≥98%, FCC
Sigma-Aldrich
1-Octanol, ≥98%, FCC, FG
Supelco
Chlorobenzene, analytical standard
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Decane, anhydrous, ≥99%
Supelco
1-Octanol, analytical standard
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Decane, ≥95%
Supelco
Decane, analytical standard
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Supelco
Decane, analytical standard
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture