Merck
Local pH tracking in living cells.
Nanoscale (2015-02-13)
Chieh-Jui Tsou, Chih-Hao Hsia, Jia-Yin Chu, Yann Hung, Yi-Ping Chen, Fan-Ching Chien, Keng C Chou, Peilin Chen, Chung-Yuan Mou
ABSTRACT
Continuous and simultaneous 3D single-particle movement and local pH detection in HeLa cells were demonstrated for the first time by combining fluorescent mesoporous silica nanoparticles (FMSNs) and a single-particle tracking (SPT) technique with a precision of ∼10 nm. FMSNs, synthesized by the co-condensation of both pH-sensitive and reference dyes with a silica/surfactant source, allow long-term reliable ratiometric pH measurements with a precision better than 0.3 pH unit because of their excellent brightness and stability. pH variation in the surrounding area of FMSNs during endocytosis was monitored in real-time. Acidification and low mobility of FMSNs were observed at the early endocytic stage, whereas basification and high mobility of FMSNs were observed at the late stage. Our results indicate that it is possible to monitor local pH changes in the environments surrounding nanoparticles during the cellular uptake process of FMSNs, which provides much needed information for designing an efficient drug delivery nanosystem.
Product Description
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Water, for TOC analysis
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, PCR Reagent
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Water, ACS reagent, for ultratrace analysis
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, Deionized
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, HPLC Plus
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)