Merck
  • Preparation and characterization of gelatin-based mucoadhesive nanocomposites as intravesical gene delivery scaffolds.
Preparation and characterization of gelatin-based mucoadhesive nanocomposites as intravesical gene delivery scaffolds.
BioMed research international (2015-01-13)
Ching-Wen Liu, Li-Ching Chang, Kai-Jen Lin, Tsan-Jung Yu, Ching-Chung Tsai, Hao-Kuang Wang, Tong-Rong Tsai
ABSTRACT
This study aimed to develop optimal gelatin-based mucoadhesive nanocomposites as scaffolds for intravesical gene delivery to the urothelium. Hydrogels were prepared by chemically crosslinking gelatin A or B with glutaraldehyde. Physicochemical and delivery properties including hydration ratio, viscosity, size, yield, thermosensitivity, and enzymatic degradation were studied, and scanning electron microscopy (SEM) was carried out. The optimal hydrogels (H), composed of 15% gelatin A175, displayed an 81.5% yield rate, 87.1% hydration ratio, 42.9 Pa·s viscosity, and 125.8 nm particle size. The crosslinking density of the hydrogels was determined by performing pronase degradation and ninhydrin assays. In vitro lentivirus (LV) release studies involving p24 capsid protein analysis in 293T cells revealed that hydrogels containing lentivirus (H-LV) had a higher cumulative release than that observed for LV alone (3.7-, 2.3-, and 2.3-fold at days 1, 3, and 5, resp.). Lentivirus from lentivector constructed green fluorescent protein (GFP) was then entrapped in hydrogels (H-LV-GFP). H-LV-GFP showed enhanced gene delivery in AY-27 cells in vitro and to rat urothelium by intravesical instillation in vivo. Cystometrogram showed mucoadhesive H-LV reduced peak micturition and threshold pressure and increased bladder compliance. In this study, we successfully developed first optimal gelatin-based mucoadhesive nanocomposites as intravesical gene delivery scaffolds.
Product Description
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
SAFC
Glycine
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Glycine, European Pharmacopoeia (EP) Reference Standard