Merck
  • Diminished viral control during simian immunodeficiency virus infection is associated with aberrant PD-1hi CD4 T cell enrichment in the lymphoid follicles of the rectal mucosa.
Diminished viral control during simian immunodeficiency virus infection is associated with aberrant PD-1hi CD4 T cell enrichment in the lymphoid follicles of the rectal mucosa.
Journal of immunology (Baltimore, Md. : 1950) (2014-09-24)
Geetha H Mylvaganam, Vijayakumar Velu, Jung-Joo Hong, Shanmugalakshmi Sadagopal, Suefen Kwa, Rahul Basu, Benton Lawson, Francois Villinger, Rama Rao Amara
ABSTRACT
The inhibitory receptor programmed death-1 (PD-1) has been shown to regulate CD8 T cell function during chronic SIV infection; however, its role on CD4 T cells, specifically in the gut-associated lymphoid tissue, is less well understood. In this study, we show that a subset of CD4 T cells expresses high levels of PD-1 (PD-1(hi)) in the rectal mucosa, a preferential site of virus replication. The majority of these PD-1(hi) CD4 T cells expressed Bcl-6 and CXCR5, markers characteristic of T follicular helper cells in the lymph nodes. Following a pathogenic SIV infection, the frequency of PD-1(hi) cells (as a percentage of CD4 T cells) dramatically increased in the rectal mucosa; however, a significant fraction of them did not express CXCR5. Furthermore, only a small fraction of PD-1(hi) cells expressed CCR5, and despite this low level of viral coreceptor expression, a significant fraction of these cells were productively infected. Interestingly, vaccinated SIV controllers did not present with this aberrant PD-1(hi) CD4 T cell enrichment, and this lack of enrichment was associated with the presence of higher frequencies of SIV-specific granzyme B(+) CD8 T cells within the lymphoid tissue, suggesting a role for antiviral CD8 T cells in limiting aberrant expansion of PD-1(hi) CD4 T cells. These results highlight the importance of developing vaccines that enhance antiviral CD8 T cells at sites of preferential viral replication and support the need for developing therapeutic interventions that limit expansion of SIV(+)PD-1(hi) CD4 T cells at mucosal sites as a means to enhance viral control.
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Propyl gallate, for microscopy, ≥98.0% (HPLC)
Sigma-Aldrich
Propyl gallate, antioxidant, ≥98.0% (HPLC)
Sigma-Aldrich
Propyl gallate, ≥98%, FCC
Sigma-Aldrich
Propyl gallate, powder
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Supelco
Propyl gallate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycerol, analytical standard