Merck
  • Lipid-lowering effects of zerumbone, a natural cyclic sesquiterpene of Zingiber zerumbet Smith, in high-fat diet-induced hyperlipidemic hamsters.
Lipid-lowering effects of zerumbone, a natural cyclic sesquiterpene of Zingiber zerumbet Smith, in high-fat diet-induced hyperlipidemic hamsters.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2014-04-09)
Thing-Fong Tzeng, Hung-Jen Lu, Shorong-Shii Liou, Chia Ju Chang, I-Min Liu
ABSTRACT
We investigated the effects of zerumbone, a natural cyclic sesquiterpene of Zingiber zerumbet Smith, on high-fat diet (HFD)-induced hyperlipidemic hamsters. After being fed HFD for 2 weeks, Syrian golden hamsters were dosed orally with zerumbone (25, 50, and 100 mg/kg) once daily for 8 weeks. Decreased plasma levels of TC, TG and LDL-C, as well as the concentrations of hepatic lipids, with a simultaneous increase in fecal lipids were found. The ratios of LDL-C/HDL-C and TC/HDL-C were elevated by zerumbone. Zerumbone exhibited the ability to decreased hepatic mRNA levels of fatty acid synthase, malic enzyme, sterol-regulatory element binding protein and 3-hydroxy-3-methyl-glutaryl-CoA reductase reductase. The hepatic mRNA expression of peroxisome proliferator-activated receptor α, together with its target gene carnitine palmitoyl transferase and acyl-CoA oxidase were also upregulated by zerumbone. Zerumbone is effective to improve dyslipidemia by modulating the genes expression involving in the lipolytic and lipogenic pathways of lipids metabolism.
Product Description
Supelco
Fructose, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
D-(−)-Fructose, analytical standard, analytical standard for fructose assay kit, for use with enzymatic assay kit FA20
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioXtra
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC)
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(−)-Fructose, tested according to Ph. Eur.
Sigma-Aldrich
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
Millipore
D-(−)-Fructose, ≥99.0% (HPLC), suitable for microbiology
Fructose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-(−)-Fructose, 98.0-102.0% dry basis, meets USP testing specifications