Merck
AAPE proliposomes for topical atopic dermatitis treatment.
Journal of microencapsulation (2014-08-05)
Alexander Jahn, Chung Kil Song, Prabagar Balakrishnan, Soon-Sun Hong, Ju-Hee Lee, Suk-Jae Chung, Dae-Duk Kim
ABSTRACT
Anti-inflammatory effect of advanced adipose stem cell derived protein extract (AAPE) could be improved by minimising protein degradation. To develop a proliposomal formulation of AAPE for the treatment of topical atopic dermatitis. Proliposomal powder was manufactured by evaporating a solution of soy phosphatidyl choline, AAPE and Poloxamer 407 in ethanol under vacuum on sorbitol powder. Characterisation of proliposomes (zeta potential, diameter, stability and flowability) as well as in vivo efficacy in a dermatitis mouse model was investigated. Reconstitution of the proliposomal powder formed liposomes of 589 ± 3.6 nm diameter with zeta potential of -51.33 ± 0.36 mV. Protein stability was maintained up to 90 days at 25 °C as proliposomes. In vivo studies on atopic dermatitis mouse model showed a significant reduction in IgE levels after topical AAPE proliposome treatment. AAPE proliposomes maintained protein stability and showed promising results for atopic dermatitis treatment.
Product Description
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), for molecular biology
Sorbitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.