Merck
  • A cyclic GB virus C derived peptide with anti-HIV-1 activity targets the fusion peptide of HIV-1.
A cyclic GB virus C derived peptide with anti-HIV-1 activity targets the fusion peptide of HIV-1.
European journal of medicinal chemistry (2014-09-15)
Ramona Galatola, Aimee Vasconcelos, Yolanda Pérez, Antonio Cruz, Montserrat Pujol, María A Alsina, María J Gómara, Isabel Haro
ABSTRACT
The development of peptide fusion inhibitors based on short synthetic peptides represents a promising option in the fight against HIV-1 infection, especially in individuals infected with multiresistant HIV-1 strains. GBV-C has the beneficial effect of retarding the progression of AIDS in people who are co-infected with both the GBV-C and HIV viruses. In previous works, the E1(22-39) GBV-C sequence (E1P8lin) was found to be capable of inhibiting the interaction of HIV-1 FP with bilayers and its cyclic analogue (E1P8cyc) showed a higher anti-HIV-1 activity. In the present work, in an attempt to gain a better understanding of the interaction of E1P8 peptides with HIV-1 FP, we analyzed direct interactions between peptides at the molecular level. Our results support that E1P8cyc might be more potent at blocking HIV-1 entry than E1P8lin as a consequence of the structure induced in the complex formed with HIV-1 FP, which is able to modify the conformation adopted by this functional domain of the HIV-1 gp41 protein in target cell membranes.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
6-(7-Nitrobenzofurazan-4-ylamino)hexanoic acid, suitable for fluorescence
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Chloroform, analytical standard
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Supelco
N,N-Dimethylformamide, analytical standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Supelco
Diethyl ether, analytical standard
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
N-Ethyldiisopropylamine, BASF quality, ≥98.0%
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
N-Ethyldiisopropylamine solution, suitable for peptide synthesis, ~2 M in 1-methyl-2-pyrrolidinone
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)