Merck
  • In vitro and in vivo biotransformation of WMS-1410, a potent GluN2B selective NMDA receptor antagonist.
In vitro and in vivo biotransformation of WMS-1410, a potent GluN2B selective NMDA receptor antagonist.
Journal of pharmaceutical and biomedical analysis (2014-02-19)
Evamaria Falck, Frank Begrow, Eugen J Verspohl, Bernhard Wünsch
ABSTRACT
Structural modification of the GluN2B selective NMDA receptor antagonist ifenprodil led to the 3-benzazepine WMS-1410 with similar GluN2B affinity but higher receptor selectivity. Herein the in vitro and in vivo biotransformation of WMS-1410 is reported. Incubation of WMS-1410 with rat liver microsomes and different cofactors resulted in four hydroxylated phase I metabolites, two phase II metabolites and five combined phase I/II metabolites. With exception of catechol 4, these metabolites were also identified in the urine of a rat treated with WMS-1410. However the metabolites 7, 8 and 12 clearly show that the catechol metabolite 4 was also formed in vivo. As shown for ifenprodil the phenol of WMS-1410 represents the metabolically most reactive structural element. The biotransformation of WMS-1410 is considerably slower than the biotransformation of ifenprodil indicating a higher metabolic stability. From the viewpoint of metabolic stability the bioisosteric replacement of the phenol of WMS-1410 by a metabolically more stable moiety should be favourable.
Sigma-Aldrich
Uridine, ≥99%
Sigma-Aldrich
Uridine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Uridine, BioUltra, ≥99%
Sigma-Aldrich
Uridine 5′-diphosphoglucuronic acid trisodium salt, 98-100%
Sigma-Aldrich
S-(5′-Adenosyl)-L-methionine iodide, ≥80% (HPLC), ≥80% (spectrophotometric assay)
Sigma-Aldrich
Uridine 5′-diphosphoglucuronic acid ammonium salt, 98-100%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%