Merck
  • Amyloid-beta-activated human microglial cells through ER-resident proteins.
Amyloid-beta-activated human microglial cells through ER-resident proteins.
Journal of proteome research (2014-11-11)
YongCheol Yoo, Kyunghee Byun, Taewook Kang, Delger Bayarsaikhan, Jin Young Kim, Seyeoun Oh, Young Hye Kim, Se-Young Kim, Won-Il Chung, Seung U Kim, Bonghee Lee, Young Mok Park
ABSTRACT
Microglial activation in the central nervous system is a key event in the neuroinflammation that accompanies neurodegenerative diseases such as Alzheimer's disease (AD). Among cytokines involved in microglial activation, amyloid β (Aβ) peptide is known to be a key molecule in the induction of diverse inflammatory products, which may lead to chronic inflammation in AD. However, proteomic studies of microglia in AD are limited due to lack of proper cell or animal model systems. In this study, we performed a proteomic analysis of Aβ-stimulated human microglial cells using SILAC (stable isotope labeling with amino acids in cell culture) combined with LC-MS/MS. Results showed that 60 proteins increased or decreased their abundance by 1.5 fold or greater. Among these, ER-resident proteins such as SERPINH1, PDIA6, PDIA3, and PPIB were revealed to be key molecular biomarkers of human microglial activation by validation of the proteomic results by immunostaining, PCR, ELISA, and Western blot. Taken together, our data suggest that ER proteins play an essential role in human microglial activation by Aβ and may be important molecular therapeutic targets for treatment of AD.
Product Description
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Trifluoroacetic acid, analytical standard
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, meets USP testing specifications
Urea, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)