Merck
  • Monitoring and removal of residual phthalate esters and pharmaceuticals in the drinking water of Kaohsiung City, Taiwan.
Monitoring and removal of residual phthalate esters and pharmaceuticals in the drinking water of Kaohsiung City, Taiwan.
Journal of hazardous materials (2014-04-08)
Gordon C C Yang, Chia-Heng Yen, Chih-Lung Wang
ABSTRACT
This study monitored the occurrence and removal efficiencies of 8 phthalate esters (PAEs) and 13 pharmaceuticals present in the drinking water of Kaohsiung City, Taiwan. The simultaneous electrocoagulation and electrofiltration (EC/EF) process was used to remove the contaminants. To this end, a monitoring program was conducted and a novel laboratory-prepared tubular carbon nanofiber/carbon/alumina composite membrane (TCCACM) was incorporated into the EC/EF treatment module (collectively designated as "TCCACM-EC/EF treatment module") to remove the abovementioned compounds from water samples. The monitoring results showed that the concentrations of PAEs were lower in water samples from drinking fountains as compared with tap water samples. No significant differences were found between the concentrations of pharmaceuticals in the two types of water samples. Under optimal operating conditions, the TCCACM-EC/EF treatment module yielded the lowest residual concentrations, ranging from not detected (ND) to 52ng/L for PAEs and pharmaceuticals of concern in the tap water samples. Moreover, the performance of the TCCACM-EC/EF treatment module is comparable with a series of treatment units employed for the drinking fountain water treatment system. The relevant removal mechanisms involved in the TCCACM-EC/EF treatment module were also discussed in this work.
Sigma-Aldrich
Benzyl butyl phthalate, 98%
Supelco
Bis(2-ethylhexyl) phthalate, PESTANAL®, analytical standard
Supelco
Benzyl butyl phthalate, analytical standard
Supelco
Dibutyl phthalate, PESTANAL®, analytical standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
1,1-Dimethylbiguanide hydrochloride, 97%
Sigma-Aldrich
Diisononyl phthalate, ester content ≥99 % (mixture of C9 isomers), technical grade
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Dibutyl phthalate, 99%
Sigma-Aldrich
Dioctyl phthalate, ≥99.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Supelco
Bis(2-ethylhexyl) phthalate, Selectophore
Supelco
Metformin hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dibutyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Benzyl butyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Bis(2-ethylhexyl) phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Melting point standard 235-237°C, analytical standard