Merck
  • Poly aspartic acid peptide-linked PLGA based nanoscale particles: potential for bone-targeting drug delivery applications.
Poly aspartic acid peptide-linked PLGA based nanoscale particles: potential for bone-targeting drug delivery applications.
International journal of pharmaceutics (2014-09-10)
Tao Jiang, Xiaohua Yu, Erica J Carbone, Clarke Nelson, Ho Man Kan, Kevin W-H Lo
ABSTRACT
Delivering drugs specifically to bone tissue is very challenging due to the architecture and structure of bone tissue. Poly(lactic-co-glycolic acid) (PLGA)-based nanoparticles (NPs) hold great promise for the delivery of therapeutics to bone tissue. The goal of the present research was to formulate a PLGA-based NP drug delivery system for bone tissue exclusively. Since poly-aspartic acids (poly-Asp) peptide sequence has been shown to bind to hydroxyapatite (HA), and has been suggested as a molecular tool for bone-targeting applications, we fabricated PLGA-based NPs linked with poly-Asp peptide sequence. Nanoparticles made of methoxy - poly(ethylene glycol) (PEG)-PLGA and maleimide-PEG-PLGA were prepared using a water-in-oil-in-water double emulsion and solvent evaporation method. Fluorescein isothiocyanate (FITC)-tagged poly-Asp peptide was conjugated to the surface of the nanoparticles via the alkylation reaction between the sulfhydryl groups at the N-terminal of the peptide and the CC double bond of maleimide at one end of the polymer chain to form thioether bonds. The conjugation of FITC-tagged poly-Asp peptide to PLGA NPs was confirmed by NMR analysis and fluorescent microscopy. The developed nanoparticle system is highly aqueous dispersible with an average particle size of ∼80 nm. In vitro binding analyses demonstrated that FITC-poly-Asp NPs were able to bind to HA gel as well as to mineralized matrices produced by human mesenchymal stem cells and mouse bone marrow stromal cells. Using a confocal microscopy technique, an ex vivo binding study of mouse major organ ground sections revealed that the FITC-poly-Asp NPs were able to bind specifically to the bone tissue. In addition, proliferation studies indicated that our FITC-poly-Asp NPs did not induce cytotoxicity to human osteoblast-like MG63 cell lines. Altogether, these promising results indicated that this nanoscale targeting system was able to bind to bone tissue specifically and might have a great potential for bone disease therapy in clinical applications.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Supelco
2-Methylbutane, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Tetramethylsilane, analytical standard, for NMR spectroscopy, ACS reagent
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tetramethylsilane, ≥99.0% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1,3-Propylene sulfite, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2-Methylbutane, anhydrous, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tetramethylsilane, ACS reagent, NMR grade, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2-Methylbutane, puriss. p.a., ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2-Methylbutane, suitable for HPLC, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dichloromethane, Selectophore, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, ACS reagent
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
SKU
Pack Size
Availability
Price
Quantity
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20