Merck
  • Increased haemodynamic adrenergic load with isoflurane anaesthesia in type 2 diabetic and obese rats in vivo.
Increased haemodynamic adrenergic load with isoflurane anaesthesia in type 2 diabetic and obese rats in vivo.
Cardiovascular diabetology (2014-12-17)
Carol T Bussey, Anne E de Leeuw, Regis R Lamberts
ABSTRACT
Increasing numbers of type 2 diabetic and obese patients with enhanced rates of cardiovascular complications require surgical interventions, however they have a higher incidence of perioperative haemodynamic complications, which has been linked to adrenergic dysfunction. Therefore, we aimed to determine how α- and β-adrenoceptor (AR)-mediated haemodynamic responses are affected by isoflurane anaesthesia in experimental type 2 diabetes and obesity in vivo. Sixteen-week old male Zucker type 2 Diabetic Fatty (ZDF) rats, Zucker Obese rats and their lean counterparts (n = 7-9 per group) were instrumented with radio telemeters to record blood pressure and heart rate and with vascular access ports for non-invasive intravenous drug delivery in vivo. Haemodynamic effects of α-AR (phenylephrine; 1-100 μg x kg(-1)) or β-AR (dobutamine; 2-120 μg x kg(-1)) stimulation were assessed under conscious and anaesthetised (isoflurane; 2%) conditions. Vascular α-AR sensitivity was increased in both diabetic (non-diabetic 80 ± 3 vs. diabetic 95 ± 4 ΔmmHg at 100 μg x kg(-1); p < 0.05) and obese (lean 65 ± 6 vs. obese 84 ± 6 ΔmmHg at 20 μg x kg(-1); p < 0.05) conscious rats. Interestingly, anaesthesia exacerbated and prolonged the increased α-AR function in both diabetic and obese animals (non-diabetic 51 ± 1 vs. diabetic 68 ± 4 ΔmmHg, lean 61 ± 5 vs. obese 84 ± 2 ΔmmHg at 20 μg x kg(-1); p < 0.05). Meanwhile, β-AR chronotropic sensitivity was reduced in conscious diabetic and obese rats (non-diabetic 58 ± 7 vs. diabetic 27 ± 8 Δbpm, lean 103 ± 12 vs. obese 61 ± 9 Δbpm at 15 μg x kg(-1); p < 0.05). Anaesthesia normalised chronotropic β-AR responses, via either a limited reduction in obese (lean 51 ± 3 vs. obese 66 ± 5 Δbpm; NS at 15 μg x kg(-1)) or increased responses in diabetic animals (non-diabetic 49 ± 8 vs. diabetic 63 ± 8 Δbpm, at 15 μg x kg(-1); NS at 15 μg x kg(-1)). Long term metabolic stress, such as during type 2 diabetes and obesity, alters α- and β-AR function, its dynamics and the interaction with isoflurane anaesthesia. During anaesthesia, enhanced α-AR sensitivity and normalised β-AR function may impair cardiovascular function in experimental type 2 diabetes and obesity.
Product Description
Supelco
Carprofen, VETRANAL®, analytical standard
Carprofen, European Pharmacopoeia (EP) Reference Standard
Supelco
Carprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Lidocaine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Trimethoprim, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Sodium chloride solution, 5 M
Lidocaine, European Pharmacopoeia (EP) Reference Standard
Prilocaine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Trimethoprim, ≥98.5%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Supelco
Trimethoprim, VETRANAL®, analytical standard
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Lidocaine, powder