Merck
  • Keratinocyte growth factor-2 intratracheal instillation significantly attenuates ventilator-induced lung injury in rats.
Keratinocyte growth factor-2 intratracheal instillation significantly attenuates ventilator-induced lung injury in rats.
Journal of cellular and molecular medicine (2014-03-22)
Jing Bi, Lin Tong, Xiaodan Zhu, Dong Yang, Chunxue Bai, Yuanlin Song, Jun She
ABSTRACT
Preservation or restoration of normal alveolar epithelial barrier function is crucial for pulmonary oedema resolution. Keratinocyte growth factor-2 (KGF-2), a potent epithelial cell mitogen, may have a role in preventing ventilator-induced lung injury (VILI), which occurs frequently in mechanically ventilated patients. The aim of the study was to test the role of KGF-2 in VILI in rats. Forty healthy adult male Sprague-Dawley rats were randomly allocated into four groups, where rats in Groups HVZP (high-volume zero positive end-expiratory pressure) and HVZP+KGF-2 were given intratracheally equal PBS and 5 mg/kg KGF-2 72 hrs before 4 hrs HVZP ventilation (20 ml/kg), respectively, while PBS and KGF-2 were administered in the same manner in Groups Control and KGF-2, which underwent tracheotomy only with spontaneous breathing. Inflammatory cytokines (tumour necrosis factor-α, macrophage inflammatory protein 2), neutrophil and total protein levels in bronchoalveolar lavage fluid and surfactant protein mRNA expression in lung tissue were detected; the number of alveolar type II cells, lung water content and lung morphology were also evaluated. The results indicate that pre-treatment with KGF-2 showed dramatic improvement in lung oedema and inflammation compared with HVZP alone, together with increased surfactant protein mRNA and alveolar type II cells. Our results suggest that KGF-2 might be considered a promising prevention for human VILI or other acute lung injury diseases.
Sigma-Aldrich
Vecuronium bromide
Vecuronium bromide, European Pharmacopoeia (EP) Reference Standard
Supelco
Urethane, analytical standard
Sigma-Aldrich
Urethane, ≥99%
Sigma-Aldrich
Urethane, ≥99.0% (GC)