Merck
  • Bicyclic peptides conjugated to an albumin-binding tag diffuse efficiently into solid tumors.
Bicyclic peptides conjugated to an albumin-binding tag diffuse efficiently into solid tumors.
Molecular cancer therapeutics (2014-11-09)
Lisa Pollaro, Sandeep Raghunathan, Julia Morales-Sanfrutos, Alessandro Angelini, Stephan Kontos, Christian Heinis
ABSTRACT
Monoclonal antibodies have long in vivo half-lives and reach high concentrations in tumors but cannot access all regions in the tissue, whereas smaller ligands such as peptides distribute better but are limited by low concentrations due to fast renal clearance. A potential solution to this problem might be offered by peptide-based ligands that are conjugated to an albumin-binding tag, and thus have a long plasma half-life. Herein, we tested if a small ligand based on a bicyclic peptide (1.9 kDa) conjugated to an albumin-binding peptide (2.3 kDa) can diffuse into tissues. Although the peptide conjugate (4.6 kDa) was most of the time bound to the large protein serum albumin (66.5 kDa), it diffused deeply into tissues and reached high nanomolar concentrations in wide areas of solid tumors. Most of the peptide conjugate isolated from tumor tissue was found to be fully intact 24 hours after administration. Because of its noncovalent interaction with albumin, the bicyclic peptide might dissociate to diffuse to tumor regions that are not accessible to larger ligands. Bicyclic peptides having high binding affinity for targets of interest and being proteolytically stable can be evolved by phage display; in conjunction with albumin-binding tags, they offer a promising format to access targets in solid tumors.
Sigma-Aldrich
Lugol solution, for microscopy (Bact., Bot.)
Sigma-Aldrich
1,4-Diazabicyclo[2.2.2]octane, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
1,3,5-Tris(bromomethyl)benzene, 97%
SAFC
HEPES
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
N-Ethyldiisopropylamine, BASF quality, ≥98.0%
Sigma-Aldrich
Thioanisole, ReagentPlus®, ≥99%
Sigma-Aldrich
N-Ethyldiisopropylamine solution, suitable for peptide synthesis, ~2 M in 1-methyl-2-pyrrolidinone
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Dabco® 33-LV
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
HEPES
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ultrapure Acetonitrile
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Iodine, ACS reagent, ≥99.8%, solid
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Iodine, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Iodine, ReagentPlus®, 99.7% trace metals basis, beads, 1-3 mm
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%