Merck
  • Expression of netrin-1 receptors in retina of oxygen-induced retinopathy in mice.
Expression of netrin-1 receptors in retina of oxygen-induced retinopathy in mice.
BMC ophthalmology (2014-08-26)
Dan Liu, Si-Qi Xiong, Lei Shang, Xiao-feng Tian, Jing Yang, Xiao-Bo Xia
ABSTRACT
Netrin-1 has been reported to promote retinal neovascularization in oxygen-induced retinopathy (OIR). However, netrin-1 receptors, which may mediate netrin-1 action during retinal neovascularization, have not been characterized. In this study, we investigated netrin-1 receptor subtype expression and associated changes in the retinas of mice with OIR. C57BL/6J mice were exposed to 75±2% oxygen for 5 days and then returned to normal air to induce retinal neovascularization. Reverse transcriptase polymerase chain reaction (RT-PCR) and Western blot were used to examine the expression of netrin-1 receptor subtypes in the mouse retinas. Double staining of netrin-1 receptor subtypes and isolectin B4 was used to determine the location of the netrin-1 receptor subtypes in the retinas. Inhibition of retinal neovascularization was achieved by UNC5B shRNA plasmid intravitreal injection. Retinal neovascularization was examined by fluorescein angiography and quantification of preretinal neovascular nuclei in retinal sections. RT-PCR results showed that, except for UNC5A, netrin-1 receptor subtypes UNC5B, UNC5C, UNC5D, DCC, neogenin, and A2b were all expressed in the retinas of OIR mice 17 days after birth. Western blots showed that only UNC5B expression was significantly increased on that day, and immunofluorescence results showed that only UNC5B and neogenin were expressed in retinal vessels. Treatment of OIR mice with the UNC5B shRNA plasmid dramatically reduced neovascular tufts and neovascular outgrowth into the inner limiting membrane. UNC5B may promote retinal neovascularization in OIR mice.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Pricing and availability is not currently available.
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Pricing and availability is not currently available.
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human UNC5B
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Pricing and availability is not currently available.
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Pricing and availability is not currently available.