Merck
  • Salt-independent hydrophobic displacement chromatography for antibody purification using cyclodextrin as supermolecular displacer.
Salt-independent hydrophobic displacement chromatography for antibody purification using cyclodextrin as supermolecular displacer.
Journal of chromatography. A (2014-12-03)
Jun Ren, Peng Yao, Jingjing Chen, Lingyun Jia
ABSTRACT
Hydrophobic interaction chromatography (HIC) offers an orthogonal selectivity to ion exchange chromatography and the combination of the two processes can provide a potential cost-effective alternative to protein A chromatography in industrial antibody purification. However, the application of HIC is limited by its close dependence on high concentrations of kosmotropic salts to achieve desired separation. These salts can cause antibody precipitation and induce the corrosion of manufacturing facilities. Here, we report a new strategy of salt-independent HIC, which can capture antibody at the physiological salt concentration and allow the recovery of bound proteins through cyclodextrin (CD)-based displacement elution. Hydrophobicity-intensified HIC media with different coupling amount of phenyl ligands were prepared and assessed for their antibody binding capacity and selectivity. β-CD was investigated for its supermolecular interaction with phenyl ligands and elution capacity as a displacer. The results clarified a nearly linear correlation between binding capacity of human immunoglobulin G (IgG) and phenyl coupling density in the range of 44-159 μmol/mL. The host-guest interaction between β-CD and the phenyl ligands revealed a modest binding strength (Ka=4.1×10(3) M(-1)), and 15 mM β-CD solution showed a general effectiveness as displacement eluent for these HIC media, with IgG recovery varying with the ligand density. This strategy allowed the direct purification of human IgG from serum with satisfactory purity. The whole procedure of this method, including loading and elution, can be performed under physiological conditions. We expect such a salt-independent mode of HIC could be used as a capture or intermediate step in industrial antibody purification.
Product Description
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Supelco
Benzoic acid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Supelco
Melting point standard 121-123°C, analytical standard
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Benzoic acid, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland