Merck
  • Auranofin is highly efficacious against Toxoplasma gondii in vitro and in an in vivo experimental model of acute toxoplasmosis.
Auranofin is highly efficacious against Toxoplasma gondii in vitro and in an in vivo experimental model of acute toxoplasmosis.
PLoS neglected tropical diseases (2014-08-01)
Rosa M Andrade, Juan D Chaparro, Edmund Capparelli, Sharon L Reed
ABSTRACT
The mainstay of toxoplasmosis treatment targets the folate biosynthetic pathways and has not changed for the last 50 years. The activity of these chemotherapeutic agents is restricted to one lifecycle stage of Toxoplasma gondii, they have significant toxicity, and the impending threat of emerging resistance to these agents makes the discovery of new therapies a priority. We now demonstrate that auranofin, an orally administered gold containing compound that was FDA approved for treatment of rheumatoid arthritis, has activity against Toxoplasma gondii in vitro (IC50 = 0.28 µM) and in vivo (1 mg/kg). Replication within human foreskin fibroblasts of RH tachyzoites was inhibited by auranofin. At 0.4 µM, auranofin inhibited replication, as measured by percent infected fibroblasts at 24 hrs, (10.94% vs. 24.66% of controls; p = 0.0003) with no effect on parasite invasion (16.95% vs. 12.91% p = 0.4331). After 18 hrs, 62% of extracellular parasites treated with auranofin were non-viable compared to control using an ATP viability assay (p = 0.0003). In vivo, a previously standardized chicken embryo model of acute toxoplasmosis was used. Fourteen day old chicken embryos were injected through the chorioallantoic vein with 1×104 tachyzoites of the virulent RH strain. The treatment group received one dose of auranofin at the time of inoculation (1 mg/kg estimated body weight). On day 5, auranofin-treated chicken embryos were 100% protected against death (p = 0.0002) and had a significantly reduced parasite load as determined by histopathology, immunohistochemistry and by the number of parasites quantified by real-time PCR. These results reveal in vitro and in vivo activity of auranofin against T. gondii, suggesting that it may be an effective alternative treatment for toxoplasmosis.
Sigma-Aldrich
Auranofin, ≥98% (HPLC)
Supelco
Pyrimethamine, VETRANAL®, analytical standard
Supelco
Sulfadiazine, VETRANAL®, analytical standard
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
Sulfadiazine, 99.0-101.0%
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Pyrimethamine, European Pharmacopoeia (EP) Reference Standard
Sulfadiazine for identification of impurity F, European Pharmacopoeia (EP) Reference Standard
Supelco
Sulfadiazine, Pharmaceutical Secondary Standard; Certified Reference Material
Sulfadiazine, European Pharmacopoeia (EP) Reference Standard