Merck
  • Identification of organic acids as potential biomarkers in the urine of autistic children using gas chromatography/mass spectrometry.
Identification of organic acids as potential biomarkers in the urine of autistic children using gas chromatography/mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-02-26)
Joanna Kałużna-Czaplińska, Ewa Żurawicz, Wiktoria Struck, Michał Markuszewski
ABSTRACT
There is a need to identify metabolic phenotypes in autism as they might each require unique approaches to prevention. Biological markers can help define autism subtypes and reveal potential therapeutic targets. The aim of the study was to identify alterations of small molecular weight compounds and to find potential biomarkers. Gas chromatography/mass spectrometry was employed to evaluate major metabolic changes in low molecular weight urine metabolites of 14 children with autism spectrum disorders vs. 10 non-autistic subjects. The results prove the usefulness of an identified set of 21 endogenous compounds (including 14 organic acids), whose levels are changed in diseased children. Gas chromatography/mass spectrometry method combined with multivariate statistical analysis techniques provide an efficient way of depicting metabolic perturbations of diseases, and may potentially be applicable as a novel strategy for the noninvasive diagnosis and treatment of autism.
Sigma-Aldrich
Chlorotrimethylsilane, puriss., ≥99.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Supelco
N-Methyl-bis(trifluoroacetamide), for GC derivatization, LiChropur, ≥99.0% (GC)
Supelco
N-Methyl-bis(trifluoroacetamide), for GC derivatization, LiChropur, ≥97.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sigma-Aldrich
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
Supelco
Chlorotrimethylsilane, for GC derivatization, LiChropur, ≥99.0% (GC)
Supelco
Pyridine, analytical standard
Sigma-Aldrich
Acetic anhydride, Arxada quality, ≥99.5% (GC)
Supelco
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, for GC derivatization, LiChropur, ≥98.5%
Supelco
N-Methyl-N-(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane, for GC derivatization, LiChropur
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Acetic anhydride, ReagentPlus®, ≥99%
Sigma-Aldrich
Chlorotrimethylsilane solution, 1.0 M in THF
Sigma-Aldrich
Acetic anhydride, ACS reagent, ≥98.0%
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Acetic anhydride, 99.5%
Supelco
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, synthesis grade
Supelco
Acetic anhydride, for GC derivatization, LiChropur, ≥99.0%
Supelco
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, BioReagent, for silylations, LiChropur
Sigma-Aldrich
Heptadecanoic acid, ≥98%
Sigma-Aldrich
Pyridine, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Supelco
Pyridine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Pyridine, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic anhydride, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99% (GC)