Merck
  • Epidermal axonal swellings in painful and painless diabetic peripheral neuropathy.
Epidermal axonal swellings in painful and painless diabetic peripheral neuropathy.
Muscle & nerve (2014-08-19)
Audrey Cheung, Peter Podgorny, Jose A Martinez, Cynthia Chan, Cory Toth
ABSTRACT
The pathophysiology of neuropathic pain (NeP) in diabetic peripheral neuropathy (DPN) is unclear. A potential pathological feature associated with intraepidermal nerve fiber density (IENFD) loss in DPN is axonal swellings. We determined the prevalence of intraepidermal axonal swellings in DPN patients with or without NeP and compared the findings with diabetes patients without DPN, patients with idiopathic neuropathy with NeP, and control subjects. The primary outcome measure was the ratio of axonal swellings to IENFD. Secondary outcome measures included clinical neuropathy severity and assessment for messenger RNA for voltage-gated sodium and calcium channels. IENFD was depressed in DPN (with/without pain) and in idiopathic neuropathy patients. Axonal swelling ratios were similar for DPN subjects with and without pain. There was no overexpression of voltage-gated ion channels in epidermis from DPN patients. Clinical neuropathy severity was only related to IENFD. There was no clinical relationship to pain or clinical neuropathy severity for axonal swellings in DPN.
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Pyridoxal 5′-phosphate hydrate, ≥98%
Sigma-Aldrich
Pyridoxal 5′-phosphate hydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Pyridoxal 5′-phosphate monohydrate, ≥97.0% (NT)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Supelco
Glycerol, analytical standard