Merck
PDGF-BB protects mitochondria from rotenone in T98G cells.
Neurotoxicity research (2014-12-18)
Ricardo Cabezas, Marcos Fidel Avila, Janneth González, Ramon Santos El-Bachá, George E Barreto
ABSTRACT
Rotenone is one of the most-studied neurotoxic substances as it induces oxidative stress processes both in cellular and animal models. Rotenone affects ATP generation, reactive oxygen species (ROS) production, and mitochondrial membrane potential in neurons and astrocyte-like cells. Previous epidemiologic studies have supported the role of neurotrophic factors such as BDNF and GDNF in neuroprotection mainly in neurons; however, only very few studies have focused on the importance of astrocytic protection in neurodegenerative models. In the present study, we assessed the neuroprotective effects of PDGF-BB against toxicity induced by rotenone in the astrocytic-like model of T98G human glioblastoma cell line. Our results demonstrated that pretreatment with PDGF-BB for 24 h increased cell viability, preserved nuclear morphology and mitochondrial membrane potential following stimulation with rotenone, and reduced ROS production nearly to control conditions. These observations were accompanied by important morphological changes induced by rotenone and that PDGF-BB was able to preserve cellular morphology under this toxic stimuli. These findings indicated that PDGF-BB protects mitochondrial functions, and may serve as a potential therapeutic strategy in rotenone-induced oxidative damage in astrocytes.
Sigma-Aldrich
Dihydroethidium, BioReagent, suitable for fluorescence, ≥95% (HPCE)
Sigma-Aldrich
Dihydroethidium, ≥95%
Sigma-Aldrich
Platelet-Derived Growth Factor-BB human, PDGF-BB, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Supelco
Dimethyl sulfoxide, analytical standard
Supelco
Rotenone, PESTANAL®, analytical standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%