Merck
  • Damaging legacy: maternal cigarette smoking has long-term consequences for male offspring fertility.
Damaging legacy: maternal cigarette smoking has long-term consequences for male offspring fertility.
Human reproduction (Oxford, England) (2014-10-02)
A P Sobinoff, J M Sutherland, E L Beckett, S J Stanger, R Johnson, A G Jarnicki, A McCluskey, J C St John, P M Hansbro, E A McLaughlin
ABSTRACT
What are the effects on fertility of cigarette smoke-induced toxicity on male offspring exposed during the gestational/weaning period? Maternal cigarette smoke exposure during the gestational/weaning period causes long-term defects in male offspring fertility. Cigarette smoke is a well-known reproductive toxicant which is particularly harmful to both fetal and neonatal germ cells. However, recent studies suggest a significant portion of young mothers in the developed world still smoke during pregnancy. In the context of male reproductive health, our understanding of the effects of in utero exposure on offspring fertility is limited. In this study, 27 C57BL/6 5-week-old female mice were exposed via the nose-only to cigarette smoke (treatment) or 27 were exposed to room air (control) for 6 weeks before being housed with stud males to produce litters. In the treatment group, smoke exposure continued throughout mating, pregnancy and lactation until weaning of pups at 21 days post birth. Male offspring were examined at post-natal days 3, 6, 12, 21 and 98 (adult). Approximately 108 maternal smoke-exposed C57BL/6 offspring and controls were examined. Spermatogenesis was examined using testicular histology and apoptosis/DNA damage was assessed using caspase immunohistochemistry and TUNEL. Sertoli cell morphology and fluctuations in the spermatogonial stem cell population were also examined using immunohistochemistry. Microarray and QPCR analysis were performed on adult testes to examine specific long-term transcriptomic alteration as a consequence of maternal smoke exposure. Sperm counts and motility, zona/oolemma binding assays, COMET analysis and mitochondrial genomic sequencing were also performed on spermatozoa obtained from adult treated and control mice. Fertility trials using exposed adult male offspring were also performed. Maternal cigarette smoke exposure caused increased gonocyte and meiotic spermatocyte apoptosis (P < 0.01) as well as germ cell depletion in the seminiferous tubules of neonatal and juvenile offspring. Aberrant testicular development characterized by abnormal Sertoli and germ cell organization, a depleted spermatogonial stem cell population (P < 0.01), atrophic seminiferous tubules and increased germ cell DNA damage (P < 0.01) persisted in adult offspring 11 weeks after exposure. Microarray analysis of adult offspring testes associated these defects with meiotic germ cell development, sex hormone metabolism, oxidative stress and Sertoli cell signalling. Next generation sequencing also revealed a high mitochondrial DNA mutational load in the testes of adult offspring (P < 0.01). Adult maternal smoke-exposed offspring also had reduced sperm counts with spermatozoa exhibiting morphological abnormalities (P < 0.01), affecting motility and fertilization potential. Odf2, a spermatozoa flagellum component required for coordinated ciliary beating, was also significantly down-regulated (P < 0.01) in maternal smoke-exposed adult offspring, with aberrant localization along the spermatozoa flagellum. Adult maternal smoke-exposed offspring took significantly longer to impregnate control females and had a slight but significant (P < 0.01) reduction in litter size. This study examined only one species (mouse) using a smoking model which only simulates human cigarette smoke exposure. This study represents the first comprehensive animal model of maternal smoking on male offspring reproductive function, suggesting that exposure during the gestational/weaning period causes long-term defects in male offspring fertility. This is due to a compromised spermatogonial stem cell population resulting from gonocyte apoptosis and impaired spermatogenic development. This results in significant germ cell damage and Sertoli cell dysfunction, impacting germ cell number, tubule organization, DNA damage and spermatozoa in adult offspring. This study strengthens the current literature suggesting that maternal exposure impairs male offspring fertility, which is currently debated due to conflicting studies. This study was funded by the Australian Research Council, Hunter Medical Research Institute, National Health and Medical Research Council of Australia and the Newcastle Permanent Building Society Charitable Trust. The authors declare no conflict of interest.
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Tyrosine, European Pharmacopoeia (EP) Reference Standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, for molecular biology, BioReagent, ≥99.5%
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
DL-Tyrosine, 99%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Monoclonal Anti-Tubulin, Tyrosine antibody produced in mouse, clone TUB-1A2, ascites fluid
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free