Merck
  • Effect of modified Chung-Sim-Youn-Ja-Tang on anti-inflammatory and anti-lipogenesis in RAW 264.7 and 3T3-L1 cells.
Effect of modified Chung-Sim-Youn-Ja-Tang on anti-inflammatory and anti-lipogenesis in RAW 264.7 and 3T3-L1 cells.
International journal of clinical and experimental pathology (2015-03-11)
Eun Hyoung Shim, Hoyoung Lee, Myeong Soo Lee
ABSTRACT
Jea-Ma-Chung-Sim-Youn-Ja-Tang (JCST), one of herbal formulae, is modified prescription base on Chung-Sim-Youn-Ja-Tang (CST) which treats cerebrovascular disease for the Tae-Eum-In (TE). This study was designed to determine the anti-inflammatory and anti-adipogenesis effect of CST and JCST in vitro. CST and JCST of various concentrations were added in RAW 264.7 and 3T3-L1 cell. To determine the anti-inflammatory and anti-adipogenesis effects of CST and JCST, the PGE2 production was measured by lipopolysaccharide (LPS) treated RAW 264.7 cell. The adipocytes was determined by Oil red O staining, triglyceride (TG) production, leptin level and the protein expressions of peroxisome proliferator-activated receptor gamma (PPARγ), CCAAT/enhancer binding proteins alpha (C/EBPα) and fatty acid binding protein 4 (FABP4) in 3T3-L1 adipocytes. Our results showed that treatment with JCST significantly decreased PGE2 production on RAW 264.7 cell and suppressed adipocyte differentiation, lipid accumulation, TG production, leptin contents and the protein expressions of PPARγ, C/EBPα and FABP4 on 3T3-L1 adipocytes compared to CST without affecting cell viability. In conclusions, our results suggest that JCST may be useful to inhibit the effect on lipid metabolism compared to CST, and regulates lipogenesis effectively. Therefore, our data provides scientific evidence to support the clinical use of JCST in the treatment of cerebrovascular diseases such as stroke in the TE.
Sigma-Aldrich
N-Chlorosuccinimide, 98%
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99%, BioUltra
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
Indomethacin, European Pharmacopoeia (EP) Reference Standard
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%