Merck
  • The effects of galectin-1 on the gene expression of the transcription factors TBX21, GATA-3, FOXP3 and RORC.
The effects of galectin-1 on the gene expression of the transcription factors TBX21, GATA-3, FOXP3 and RORC.
Molecular and cellular biochemistry (2014-10-09)
V D Yakushina, O A Vasil'eva, N V Ryazantseva, V V Novitsky, L A Tashireva
ABSTRACT
CD4(+) T cells orchestrate the immune response by differentiating into T helper (Th) or regulatory (Treg) cell subsets that secrete distinct sets of cytokines. They also play a critical role in the pathogenesis of autoimmunity, asthma, allergy and, likely, cancer. The mechanisms involved in the regulation of CD4(+) T cell homeostasis by galectin-1 remain poorly characterized. To investigate whether galectin-1 modulates the differentiation of CD4(+) T cells, the effects of galectin-1 on the mRNA expression levels of TBX21, GATA-3, FOXP3 and RORC in activated peripheral blood mononuclear cells were examined. The expression levels of GATA-3 and FOXP3 mRNA were up-regulated after treatment with 1.0 μg/ml galectin-1 and were unchanged (for GATA-3) or slightly elevated (for FOXP3) compared with untreated cells when 2.0 μg/ml galectin-1 was added. At the same time, at both concentrations of galectin-1, we observed reduced TBX21 and RORC mRNA expression levels. These findings support the concept that galectin-1 skews the differentiation of CD4(+) T cells towards Th2 and Treg cells.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis