Merck
  • Development of a novel calcium phosphate cement composed mainly of calcium sodium phosphate with high osteoconductivity.
Development of a novel calcium phosphate cement composed mainly of calcium sodium phosphate with high osteoconductivity.
Journal of materials science. Materials in medicine (2014-03-29)
Masashi Tanaka, Mitsuru Takemoto, Shunsuke Fujibayashi, Toshiyuki Kawai, Masako Tsukanaka, Kimiaki Takami, Satoshi Motojima, Hikaru Inoue, Takashi Nakamura, Shuichi Matsuda
ABSTRACT
Two novel calcium phosphate cements (CPC) have been developed using calcium sodium phosphate (CSP) as the main ingredient. The first of these cements, labeled CAC, contained CSP, α-tricalcium phosphate (TCP), and anhydrous citric acid, whereas the second, labeled CABC, contained CSP, α-TCP, β-TCP, and anhydrous citric acid. Biopex(®)-R (PENTAX, Tokyo, Japan), which is a commercially available CPC (Com-CPC), and OSferion(®) (Olympus Terumo Biomaterials Corp., Tokyo, Japan), which is a commercially available porous β-TCP, were used as reference controls for analysis. In vitro analysis showed that CABC set in 5.7 ± 0.3 min at 22 °C and had a compressive strength of 86.0 ± 9.7 MPa after 5 days. Furthermore, this material had a compressive strength of 26.7 ± 3.7 MPa after 2 h in physiologic saline. CAC showed a statistically significantly lower compressive strength in the presence of physiologic saline and statistically significantly longer setting times than those of CABC. CABC and CAC exhibited apatite-forming abilities in simulated body fluid that were faster than that of Com-CPC. Samples of the materials were implanted into the femoral condyles of rabbits for in vivo analysis, and subsequent histological examinations revealed that CABC exhibited superior osteoconductivity and equivalent bioresorbability compared with Com-CPC, as well as superior osteoconductivity and bioresorbability compared with CAC. CABC could therefore be used as an alternative bone substitute material.
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Calcium carbonate, ACS reagent, chelometric standard, 99.95-100.05% dry basis
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Calcium carbonate, tested according to Ph. Eur.
Supelco
Calcium carbonate, reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Calcium carbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
Supelco
Glycerol, analytical standard
Supelco
Calcium carbonate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Calcium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Calcium carbonate, ReagentPlus®
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Calcium carbonate, ACS reagent, ≥99.0%, powder