Merck
  • The potential of neurotensin secreted from neuroendocrine tumor cells to promote gelsolin-mediated invasiveness of prostate adenocarcinoma cells.
The potential of neurotensin secreted from neuroendocrine tumor cells to promote gelsolin-mediated invasiveness of prostate adenocarcinoma cells.
Laboratory investigation; a journal of technical methods and pathology (2015-01-13)
Kohei Hashimoto, Yuki Kyoda, Toshiaki Tanaka, Toshihiro Maeda, Ko Kobayashi, Kohsuke Uchida, Hiroshi Kitamura, Koichi Hirata, Taiji Tsukamoto, Naoya Masumori
ABSTRACT
Neuroendocrine (NE) cells in prostate cancer have been shown to be associated with the progression of prostate cancer. However, little is known about the molecular basis of this association. We have previously demonstrated that NE cells promote metastasis of a human prostate cancer cell line (LNCaP) with overexpression of the gelsolin gene. The purpose of this study was to investigate the interactions between NE cells and LNCaP cells and the involvement of gelsolin in contributing to the invasive potential of LNCaP cells. In addition, we examined whether neurotensin induced gelsolin-mediated invasion. We used the NE cell line NE-CS that was established from the prostate of the LPB-Tag 12T-10 transgenic mouse. Small interfering RNA (siRNA) targeting gelsolin or not targeting it was transfected into LNCaP cells. Cell invasion was assessed by Matrigel invasion assay. The supernatant of NE-CS cells and neurotensin induced the transformation of LNCaP cells. Neurotensin was observed in the supernatant of NE-CS cells but not in LNCaP cells. The siRNA targeting of gelsolin resulted in inhibition of invasion of LNCaP cells in the culture medium with neurotensin added, and in the supernatant of NE-CS cells with epidermal growth factor. The invasive potential of LNCaP cells enhanced by neurotensin or the supernatant of NE-CS cells through neurotensin receptor 1 (NTSR1) was blocked by a phospholipase Cγ inhibitor and an intracellular calcium chelator, with concomitant gelsolin suppression. This study indicates that NE cells and neurotensin induce gelsolin-mediated invasion of LNCaP cells through NTSR1 activation.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, suitable for enzyme immunoassay, ≥99.0% (enzymatic)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Phosphatase substrate, 5 mg tablets
Sigma-Aldrich
Phosphatase substrate, 40 mg tablets
Sigma-Aldrich
Phosphatase substrate, powder
Sigma-Aldrich
Phosphatase substrate, 100 mg capsules
Sigma-Aldrich
Phosphatase substrate, 40 mg capsules
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Phosphatase substrate, Suitable for manufacturing of diagnostic kits and reagents
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%