Merck
  • Brinzolamide nanocrystal formulations for ophthalmic delivery: reduction of elevated intraocular pressure in vivo.
Brinzolamide nanocrystal formulations for ophthalmic delivery: reduction of elevated intraocular pressure in vivo.
International journal of pharmaceutics (2014-04-01)
Annika Tuomela, Peng Liu, Jooseppi Puranen, Seppo Rönkkö, Timo Laaksonen, Giedrius Kalesnykas, Olli Oksala, Jukka Ilkka, Johanna Laru, Kristiina Järvinen, Jouni Hirvonen, Leena Peltonen
ABSTRACT
Nanocrystal-based drug delivery systems provide important tools for ocular formulation development, especially when considering poorly soluble drugs. The objective of the study was to formulate ophthalmic, intraocular pressure (IOP) reducing, nanocrystal suspensions from a poorly soluble drug, brinzolamide (BRA), using a rapid wet milling technique, and to investigate their IOP reducing effect in vivo. Different stabilizers for the nanocrystals were screened (hydroxypropyl methylcellulose (HPMC), poloxamer F127 and F68, polysorbate 80) and HPMC was found to be the only successful stabilizer. In order to investigate both the effect of an added absorption enhancer (polysorbate 80) and the impact of the free drug in the nanocrystal suspension, formulations in phosphate buffered saline (PBS) at pH 7.4 and pH 4.5 were prepared. Particle size, polydispersity (PI), solid state (DSC), morphology (SEM) as well as dissolution behavior and the uniformity of the formulations were characterized. There was rapid dissolution of BRA (in PBS pH 7.4) from all the nanocrystal formulations; after 1 min 100% of the drug was fully dissolved. The effect was significantly pronounced at pH 4.5, where the dissolved fraction of drug was the highest. The cytotoxicity of nanocrystal formulations to human corneal epithelial cell (HCE-T) viability was tested. The effects of the nanocrystal formulations and the commercial product on the cell viability were comparable. The intraocular pressure (IOP) lowering effect was investigated in vivo using a modern rat ocular hypertensive model and elevated IOP reduction was seen in vivo with all the formulations. Notably, the reduction achieved in experimentally elevated IOP was comparable to that obtained with a marketed product. In conclusion, various BRA nanocrystal formulations, which all showed advantageous dissolution and absorption behavior, were successfully formulated.
Product Description
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, BioXtra
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, ≥98%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material