Merck
  • Analysis of extensively washed hair from cocaine users and drug chemists to establish new reporting criteria.
Analysis of extensively washed hair from cocaine users and drug chemists to establish new reporting criteria.
Journal of analytical toxicology (2014-08-08)
Cynthia L Morris-Kukoski, Madeline A Montgomery, Rena L Hammer
ABSTRACT
Samples from a self-proclaimed cocaine (COC) user, from 19 drug users (postmortem) and from 27 drug chemists were extensively washed and analyzed for COC, benzoylecgonine, norcocaine (NC), cocaethylene (CE) and aryl hydroxycocaines by liquid chromatography-tandem mass spectrometry. Published wash criteria and cutoffs were applied to the results. Additionally, the data were used to formulate new reporting criteria and interpretation guidelines for forensic casework. Applying the wash and reporting criteria, hair that was externally contaminated with COC was distinguished from hair collected from individuals known to have consumed COC. In addition, CE, NC and hydroxycocaine metabolites were only present in COC users' hair and not in drug chemists' hair. When properly applied, the use of an extended wash, along with the reporting criteria defined here, will exclude false-positive results from environmental contact with COC.
Product Description
Supelco
DL-Dithiothreitol solution, 1 M in H2O
SAFC
BIS-TRIS
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
SAFC
BIS-TRIS
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium cholate hydrate, BioXtra, ≥99%
Sigma-Aldrich
Sodium cholate hydrate, suitable for cell culture, BioReagent
Sigma-Aldrich
Sodium cholate hydrate, from bovine and/or ovine bile, ≥99%
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium cholate hydrate, ≥97.0% (dried material, NT)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ≥99.0%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%