Merck
  • Oral administration of fermented probiotics improves the condition of feces in adult horses.
Oral administration of fermented probiotics improves the condition of feces in adult horses.
Journal of equine science (2015-01-06)
Saori Ishizaka, Akira Matsuda, Yosuke Amagai, Kumiko Oida, Hyosun Jang, Yuko Ueda, Masaki Takai, Akane Tanaka, Hiroshi Matsuda
ABSTRACT
The effects of probiotics on horses are still controversial. The present study was a randomized, double-blinded, placebo-controlled crossover study designed to evaluate the ability of probiotics to improve intestinal conditions in adult horses. Fermented probiotics were administered to 10 healthy adult geldings for 28 days. The clinical condition of the horses was monitored daily, and the blood and feces were biochemically analyzed every 14 days. In the probiotic-treated group, the concentration of carboxylic acids in the feces was increased at days 14 and 28. In contrast to the fecal pH in the control group, which increased at days 14 and 28, the fecal pH in the probiotic-treated group did not increase. Additionally, the relative amounts of enteropathogenic bacterial DNA were diminished in the probiotic-treated group. These results suggest that probiotic bacteria proliferated in the equine intestine. No instances of abnormal clinical conditions or abnormal values in blood tests were observed throughout the study. Oral administration of fermented probiotics may have the ability to improve the intestinal environment biochemically and microbiologically without the risk of adverse effects.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Skatole, ≥98%
Sigma-Aldrich
4-Ethylphenol, ≥97.0% (GC)
Supelco
p-Cresol, analytical standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Phenol-d6, 99 atom % D
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
4-Ethylphenol, 99%
Sigma-Aldrich
Indole, ≥99%, FG
Sigma-Aldrich
4-Ethylphenol, ≥98%, FG
Sigma-Aldrich
Indole, ≥99%
Sigma-Aldrich
p-Cresol, ≥99%, FG
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
p-Cresol, 99%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.