Merck
  • Characteristics of titanium dioxide microdispersions with different photo-activity suitable for sunscreen formulations.
Characteristics of titanium dioxide microdispersions with different photo-activity suitable for sunscreen formulations.
International journal of pharmaceutics (2015-02-07)
L Kubáč, J Akrman, K Kejlová, H Bendová, K Klánová, Z Hladíková, P Pikal, L Kovaříková, L Kašparová, D Jírová
ABSTRACT
The aim of the study was the comparison of photo-activity of three types of titanium dioxide (TiO2) micro-dispersions intended for use as UV filters for cosmetic sunscreen products. The dispersions were also investigated with regard to their influence on the stability of photo-protective systems in cosmetic emulsions, their skin penetration/absorption and their photo-toxicity for humans and skin bacterial flora. All the tested micro-dispersions of rutile TiO2 type (agglomerates with diameter 120-150 nm), with primary particle size lower than 100 nm, demonstrated no phototoxic effect and insignificant antimicrobial behaviour. On the other hand, TiO2 with insufficient deactivation of photo-activity had significant negative impact on the stability of other organic UV filters and therefore on the stability of declared UV protective factors (SPF, UVA-PF). The study demonstrated that the level of deactivation of TiO2 is one of the highly important factors for evaluation of UV filters used as sunscreens.
Sigma-Aldrich
Isopropyl myristate, ≥98%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Isopropyl myristate, 98%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Isopropyl myristate, ≥90% (GC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
Supelco
Isopropyl myristate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
SAFC
Sodium chloride solution, 5 M