Merck
  • Comparative functional characterization of a novel benzoate hydroxylase cytochrome P450 of Fusarium oxysporum.
Comparative functional characterization of a novel benzoate hydroxylase cytochrome P450 of Fusarium oxysporum.
Enzyme and microbial technology (2015-02-11)
Pradeepraj Durairaj, Eunok Jung, Hyun Ho Park, Byung-Gee Kim, Hyungdon Yun
ABSTRACT
FoCYP53A19, a novel cytochrome P450 capable of performing benzoate hydroxylation, was identified and characterized from the ascomycete Fusarium oxysporum f.sp. lycopersici. Comparative functional analysis of FoCYP53A19 with the heterologous and homologous cytochrome P450 reductases (CPR) such as Saccharomyces cerevisiae (ScCPR), Candida albicans (CaCPR) and F. oxysporum (FoCPR) revealed novel catalytic properties. The catalytic efficiency and substrate specificity of FoCYP53A19 were significantly influenced and altered by the source of the reductase employed. The yeast reconstitution system of FoCYP53A19 with ScCPR performed the hydroxylation of benzoic acid (BA) and demethylation of 3-methoxybenzoic acid (3-MBA); but when reconstituted with CaCPR, FoCYP53A19 performed only the essential hydroxylation of fungal benzoate catabolism. Remarkably, FoCYP53A19 with its homologous reductase FoCPR, not only demonstrated the improved conversion rates of BA and 3-MBA, but also exhibited activity toward the hydroxylation of 3-hydroxybenzoic acid. The electron transfer compatibility and the coupling efficiency between the homologous FoCYP-FoCPR system are significant and it favored enhanced monooxygenase activity with broader substrate specificity.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
3-Methoxybenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Yeast from Saccharomyces cerevisiae, Type II
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Supelco
Melting point standard 121-123°C, analytical standard
Supelco
4-Hydroxybenzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Benzoic acid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
3-Hydroxybenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Supelco
3,4-Dihydroxybenzoic acid, analytical standard