Merck
Complex influence of dermatan sulphate on breast cancer cells.
Experimental biology and medicine (Maywood, N.J.) (2014-06-11)
Ewa M Koźma, Grzegorz Wisowski, Małgorzata Latocha, Damian Kusz, Krystyna Olczyk
ABSTRACT
Tumor transformation and progression both lead to extracellular matrix remodeling, which is also reflected in an alteration in the proportion of dermatan sulphate (DS) and chondroitin sulphate (CS) and an accumulation of the latter. In addition, a significant increase in the 6-O-sulphated disaccharide contribution to the structure of both glycosaminoglycans has been observed. It is commonly accepted that CS is more permissive for tumor growth than DS. However, the detailed role of DS in tumor progression is poorly known. We tested the effects of structurally different DSs on the behavior of cultured breast cancer cells. At a high dose (10 µg/mL), all of the DSs significantly reduced cancer cell growth, although some differences in the efficiency of action were apparent. In contrast, when used at a concentration of 1 µg/mL, the examined DSs evoked different responses ranging from the stimulation to the inhibition of cancer cell proliferation. The highest stimulatory activity was associated with fibrosis-affected fascia decorin DS, which is characterized by a particularly high content of 6-O-sulphated disaccharides. Further reduction in DS concentration to 0.5 µg/mL preserved majority of biological effects which were apparent at a dose of 1 µg/mL. The enzymatic fragmentation of the DSs, particularly by chondroitinase AC I, abolished the impact exerted by 1 µg/mL of the intact DS chains and sometimes resulted in the opposite effect. In contrast to DSs, highly sulphated C-6-S exhibited no effect on the cancer cells. Our data revealed the complexity of the effects of DSs on breast cancer cells, which include both co-receptor activity and the prevention of vascular endothelial growth factor action. In addition, the biological effect of DSs is strongly dependent not only on the glycosaminoglycan structure but also on its content in the cancer environment.
Product Description
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
Sodium cyanoborohydride solution, 1.0 M in THF
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Guanidine hydrochloride solution, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Sodium cyanoborohydride solution, 5.0 M in 1 M NaOH
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Sigma-Aldrich
Guanidine hydrochloride solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Trometamol, European Pharmacopoeia (EP) Reference Standard