Merck
  • Suppression of nuclear factor-kappa B and mitogen-activated protein kinase signalling pathways by goshonoside-F5 extracted from Rubi Fructus.
Suppression of nuclear factor-kappa B and mitogen-activated protein kinase signalling pathways by goshonoside-F5 extracted from Rubi Fructus.
International immunopharmacology (2014-12-20)
Jian-Ming He, Shi-Cai Chen, Run-Ping Li, Liang-Xi Yuan, Jun-Min Bao, Mei-Li Guo
ABSTRACT
Rubi Fructus, a traditional Chinese medicine, was considered as an anti-inflammatory agent in folk medicine. In the present study, we investigated the signalling pathways involved in the anti-inflammatory effects of goshonoside-F5 (GF5), isolated from Rubi Fructus, in peritoneal macrophages and examined its therapeutic effect in a mouse endotoxic shock model. GF5 decreased NO and PGE2 production in LPS-stimulated macrophages (IC50=3.84 and 3.16μM). This effect involved the suppression of NOS-2 and COX-2 gene expression at the transcriptional level. Examination of the effects of GF5 on NF-κB signalling demonstrated that it inhibits the phosphorylation of IκB-α and IκB-β, blocking their degradation and the nuclear translocation of the NF-κB p65 subunit. Moreover, inhibition of MAPK signalling was also observed, and phosphorylation of p38 and JNK was suppressed in the presence of GF5. Inflammatory cytokines, including IL-6 and TNF-α, were down-regulated by this compound after activation with LPS (IC50=17.04 and 4.09μM). Additionally, GF5 (30 and 90mg/kg, i.p.) significantly reduced the circulating cytokine levels (IL-6 and TNF-α) and increased survival in a mouse model of endotoxemia. These results show that GF5 significantly inhibits the pro-inflammatory response induced by LPS, both in vitro and in vivo. Our results provide a strong pharmacological basis for further understanding the potential therapeutic role of GF5 in inflammatory disease and shed new light on the bioactivity of ent-labdane diterpene glucoside.
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
Supelco
Tetramethylsilane, analytical standard, for NMR spectroscopy, ACS reagent
Sigma-Aldrich
Tetramethylsilane, ≥99.0% (GC)
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Sigma-Aldrich
Tetramethylsilane, ACS reagent, NMR grade, ≥99.9%
Supelco
Acetonitrile, analytical standard
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)