Merck
  • The preliminary study of p53 codon 72 polymorphism and risk of cervical carcinoma in Gabonese women.
The preliminary study of p53 codon 72 polymorphism and risk of cervical carcinoma in Gabonese women.
Medical oncology (Northwood, London, England) (2014-12-17)
Samira Zoa Assoumou, Anicet Luc M Boumba, Angelique Ndjoyi-Mbiguino, Abdelkrim Khattabi, Moulay Mustapha Ennaji
ABSTRACT
Cervical cancer is the leading cause of cancer-related death in Africa and the first most common cancer in Gabonese women due to infection of high-risk human papillomavirus (HPV). However, other cofactors such as genetic factors also come into play. A common polymorphism of the p53 codon 72 in exon 4 with two alleles encoding arginine or proline is known at this locus. The homozygous arginine form of this polymorphism has been associated with the development of cervical cancer as an increased genetic risk factor. However, the results are still controversial. This study aims to investigate whether the genotype distribution of p53 codon 72 may be a risk factor for cervical cancer among Gabonese women. Samples from 102 Gabonese women, 31 diagnosed with cervical cancer and 71 healthy controls, were used. HPV detection was done by nested PCR with MY09/11 and GP5+/6+ primers followed by sequencing for HPV genotyping. p53 codon 72 polymorphism determination was performed by allele-specific PCR assay. Viral DNA was detected in 87.1 % of cases and in 54.93 % of control. HPV 16 was the most predominant in cancer and controls cases. The distribution of Arg/Arg, Arg/Pro and Pro/Pro genotypes was 35.5, 51.6 and 12.9 % in the cervical cancer group and 22.5, 62 and 15.5 % in the control group. No significant association was found between polymorphism of p53 itself as well as in combination with HPV16/18 infection and risk of development of cervical cancer among Gabonese women. Thus, the polymorphism of p53 codon 72 in exon 4 does not seem to play a role in the development of cervical cancer among Gabonese women.
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Sigma-Aldrich
Ethidium bromide, BioReagent, for molecular biology, powder
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)