Merck
  • Ethanol attenuation of long-term depression in the nucleus accumbens can be overcome by activation of TRPV1 receptors.
Ethanol attenuation of long-term depression in the nucleus accumbens can be overcome by activation of TRPV1 receptors.
Alcoholism, clinical and experimental research (2014-11-26)
Rafael Renteria, Zachary M Jeanes, Richard A Morrisett
ABSTRACT
Altered expression of synaptic plasticity within the nucleus accumbens (NAc) constitutes a critical neuroadaptive response to ethanol (EtOH) and other drugs of abuse. We have previously reported that N-methyl-D-aspartate receptor (NMDAR)-dependent long-term depression (LTD) is markedly affected by chronic intermittent ethanol exposure in vivo; however, endocannabinoid (eCB)-dependent synaptic depression, despite being very well-documented in the dorsal striatum, is much less well understood in the NAc. Whole cell patch clamp electrophysiology was used to investigate interactions between these different plasticity-induction systems. Excitatory postsynaptic currents (EPSCs) were measured in the NAc shell and NMDAR-LTD was induced by a pairing protocol (500 stimuli at 1 Hz stimulation [low-frequency stimulation (LFS)] paired with postsynaptic depolarization to -50 mV). AM251, a CB1 receptor antagonist, was used to determine whether this form of LTD is modulated by eCBs. To determine the effect of EtOH on a purely eCB-dependent response in the NAc, depolarization-induced suppression of excitation (DSE) was used in the presence of 40 mM EtOH. Finally, we determined whether the enhancement of eCB signaling with URB597, a fatty acid amide hydrolase inhibitor, and AM404, an anandamide re-uptake inhibitor would also modulate LFS LTD in the presence of NMDAR blockade or EtOH. In the presence of AM251, the LFS pairing protocol resulted in NMDAR-dependent long-term potentiation that was blocked with either EtOH or DL-APV. We also found that DSE in the NAc shell was blocked by AM251 and suppressed by EtOH. Enhanced eCB signaling rescued NAc-LTD expression in the presence of EtOH through a distinct mechanism requiring activation of TRPV1 receptors. EtOH modulation of synaptic plasticity in the NAc is dependent upon a complex interplay between NMDARs, eCBs, and TRPV1 receptors. These findings demonstrate a novel form of TRPV1-dependent LTD in the NAc shell that may be critical for EtOH dependence.
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
SAFC
Sodium chloride solution, 5 M
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C