Merck
  • Temperature dependence of production of structured triacylglycerols in the alga Trachydiscus minutus.
Temperature dependence of production of structured triacylglycerols in the alga Trachydiscus minutus.
Phytochemistry (2015-01-08)
Tomáš Řezanka, Jaromír Lukavský, Karel Sigler, Linda Nedbalová, Milada Vítová
ABSTRACT
This study describes the identification of regioisomers and enantiomers of triacylglycerols of C20 polyunsaturated fatty acids (PUFAs) in the alga Trachydiscus minutus cultivated at different temperatures using reversed- and chiral-phase liquid chromatography-mass spectrometry. The use of the two different phases contributes to ready identification, both qualitative and semiquantitative, of regioisomers and enantiomers of triacylglycerols containing eicosapentaenoic and arachidonic in the molecule. The ratio of regioisomers and enantiomers of triacylglycerols (TAG) depends on the temperature of cultivation; with lowering temperature the proportion of the achiral TAG increases and the enantiomer ratio diverges from 1:1.
Product Description
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Supelco
Tetrahydrofuran, analytical standard
Supelco
Methanol, analytical standard
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Dess-Martin periodinane, 97%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
1,2-Dimyristoyl-sn-glycero-3-phosphocholine, ≥99%
Sigma-Aldrich
Methanol, NMR reference standard
Valaciclovir impurity G, European Pharmacopoeia (EP) Reference Standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 250 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, ReagentPlus®, ≥99.0%, contains 250 ppm BHT as inhibitor
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%