Merck
  • Intravitreal ranibizumab for patients with neovascular age-related macular degeneration with good baseline visual acuity.
Intravitreal ranibizumab for patients with neovascular age-related macular degeneration with good baseline visual acuity.
Ophthalmologica. Journal international d'ophtalmologie. International journal of ophthalmology. Zeitschrift fur Augenheilkunde (2014-11-22)
Aki Kato, Tsutomu Yasukawa, Keiji Suga, Yoshio Hirano, Miho Nozaki, Munenori Yoshida, Yuichiro Ogura
ABSTRACT
To report the 1-year results of intravitreal ranibizumab (IVR) injections for neovascular age-related macular degeneration (nAMD) in patients with good baseline visual acuity (VA). Thirty-six eyes of 36 patients with nAMD with best-corrected VAs (BCVAs) >0.6 (equal to 0.22 in the logarithm of the minimum angle of resolution unit) were enrolled. IVR was the primary treatment; additional treatment was administered as needed. BCVAs and central retinal thickness (CRT) were measured periodically. The mean number of injections at month 12 was 3.3. The mean BCVAs were 0.11 ± 0.02 at baseline and 0.12 ± 0.03 at month 12, which did not significantly differ. The mean CRT significantly improved from 320 ± 15 to 254 ± 12 μm at month 12 (p < 0.01). Photodynamic therapy was applied in 2 cases because of frequent recurrences. IVR maintained VA and improved morphological changes in wet AMD with good baseline VA.
Sigma-Aldrich
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine, ≥99%
Sigma-Aldrich
Triamcinolone