Merck
  • Microbial Synthesis of the Forskolin Precursor Manoyl Oxide in an Enantiomerically Pure Form.
Microbial Synthesis of the Forskolin Precursor Manoyl Oxide in an Enantiomerically Pure Form.
Applied and environmental microbiology (2014-09-23)
Morten T Nielsen, Johan Andersen Ranberg, Ulla Christensen, Hanne Bjerre Christensen, Scott J Harrison, Carl Erik Olsen, Björn Hamberger, Birger Lindberg Møller, Morten H H Nørholm
ABSTRACT
Forskolin is a promising medicinal compound belonging to a plethora of specialized plant metabolites that constitute a rich source of bioactive high-value compounds. A major obstacle for exploitation of plant metabolites is that they often are produced in small amounts and in plants difficult to cultivate. This may result in insufficient and unreliable supply leading to fluctuating and high sales prices. Hence, substantial efforts and resources have been invested in developing sustainable and reliable supply routes based on microbial cell factories. Here, we report microbial synthesis of (13R)-manoyl oxide, a proposed intermediate in the biosynthesis of forskolin and other medically important labdane-type terpenoids. Process optimization enabled synthesis of enantiomerically pure (13R)-manoyl oxide as the sole metabolite, providing a pure compound in just two steps with a yield of 10 mg/liter. The work presented here demonstrates the value of a standardized bioengineering pipeline and the large potential of microbial cell factories as sources for sustainable synthesis of complex biochemicals.
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Supelco
Hexane, analytical standard
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Geranylgeranyl pyrophosphate ammonium salt, solution, ≥95% (TLC), ~1 mg/mL in methanol: NH4OH (7:3)
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%