Merck
  • Fractionation and identification of minor and aroma-active constituents in Kangra orthodox black tea.
Fractionation and identification of minor and aroma-active constituents in Kangra orthodox black tea.
Food chemistry (2014-08-26)
Robin Joshi, Ashu Gulati
ABSTRACT
The aroma constituents of Kangra orthodox black tea were isolated by simultaneous distillation extraction (SDE), supercritical fluid extraction and beverage method. The aroma-active compounds were identified using gas chromatography-olfactometry-mass spectrometry. Geraniol, linalool, (Z/E)-linalool oxides, (E)-2-hexenal, phytol, β-ionone, hotrienol, methylpyrazine and methyl salicylate were major volatile constituents in all the extracts. Minor volatile compounds in all the extracts were 2-ethyl-5-methylpyrazine, ethylpyrazine, 2-6,10,14-trimethyl-2-pentadecanone, acetylfuran, hexanoic acid, dihydroactinidiolide and (E/Z)-2,6-nonadienal. The concentrated SDE extract was fractionated into acidic, basic, water-soluble and neutral fractions. The neutral fraction was further chromatographed on a packed silica gel column eluted with pentane and diethyl ether to separate minor compounds. The aroma-active compounds identified using gas chromatography-olfactometry-mass spectrometry were 2-amylfuran, (E/Z)-2,6-nonadienal, 1-pentanol, epoxylinalool, (Z)-jasmone, 2-acetylpyrrole, farnesyl acetone, geranyl acetone, cadinol, cubenol and dihydroactinidiolide. AEDA studies showed 2-hexenal, 3-hexenol, ethylpyrazine, (Z/E)-linalool oxides, linalool, (E/Z)-2,6-nonadienal, geraniol, phenylethanol, β-ionone, hotrienol and dihydroactinidiolide to be odour active components.
Product Description
Supelco
(−)-Linalool, analytical standard
Sigma-Aldrich
(−)-Linalool, ≥95.0% (sum of enantiomers, GC)
Sigma-Aldrich
Nerolidol, mixture of cis and trans, ≥97%, stabilized, FG
Sigma-Aldrich
cis-3,7-Dimethyl-2,6-octadien-1-ol, 97%
Sigma-Aldrich
Nerolidol, 98%, Mixture of cis and trans
Sigma-Aldrich
cis-3,7-Dimethyl-2,6-octadien-1-ol, ≥97%, FCC, FG
Supelco
Nerol, analytical standard
Sigma-Aldrich
Methyl salicylate, tested according to Ph. Eur.
Supelco
Geraniol, analytical standard
Sigma-Aldrich
Linalool, 97%
Sigma-Aldrich
1-Phenylethanol, 98%
Sigma-Aldrich
α-Methylbenzyl alcohol, ≥99%, FCC, FG
Sigma-Aldrich
Methyl salicylate, ≥98%, FCC, FG
Sigma-Aldrich
Geraniol, 98%
Sigma-Aldrich
Linalool, ≥97%, FCC, FG
Sigma-Aldrich
Geraniol, ≥97%, FCC, FG
Sigma-Aldrich
Methyl salicylate, ReagentPlus®, ≥99% (GC)
Supelco
Methyl salicylate, analytical standard
Supelco
Linalool, analytical standard
Sigma-Aldrich
Geraniol, natural, ≥97%, FG
Supelco
Methyl salicylate, Pharmaceutical Secondary Standard; Certified Reference Material