Merck
  • A NOS3 polymorphism determines endothelial response to folate in children with type 1 diabetes or obesity.
A NOS3 polymorphism determines endothelial response to folate in children with type 1 diabetes or obesity.
The Journal of pediatrics (2014-12-06)
Esko J Wiltshire, Alexia S Peña, Karen MacKenzie, Tulika Bose-Sundernathan, Roger Gent, Jennifer J Couper
ABSTRACT
To determine the effect of polymorphisms in NOS3 and folate pathway enzymes on vascular function and folate status and endothelial response to folate in children with diabetes or obesity. A total of 244 subjects (age 13.8 ± 2.8 years, 125 males) were studied for NOS3 and/or folate pathway polymorphisms using polymerase chain reaction/restriction fragment length polymorphism, including at baseline: 139 with type 1 diabetes; 58 with obesity; and 47 controls. The effect of NOS3 genotype on endothelial response to folate (5 mg) was assessed in 85 subjects with diabetes and 28 obese subjects who received active treatment during intervention trials. Vascular function (flow-mediated dilatation [FMD] and glyceryl trinitrate-mediated dilatation), clinical, and biochemical measurements were assessed at baseline and 8 weeks in folate intervention studies. Folate pathway enzyme and NOS3 polymorphisms did not significantly affect baseline vascular function. The polymorphism in intron 4 of endothelial nitric oxide synthase altered endothelial response to folate significantly: in subjects with diabetes FMD improved by 6.4 ± 5% (insertion carriers) vs 2.3 ± 6.6% (deletion carriers), P = .01; in obese subjects FMD improved by 1.8 ± 5.4% (insertion carriers) and deteriorated by -3.2 ± 7.2% (deletion carriers), P = .05. More subjects carrying the insertion normalized FMD after folate supplementation (insertion 64% vs deletion 28%, χ(2) = 10.14, P = .001). A NOS3 polymorphism predicts endothelial response to folate in children with diabetes or obesity, with implications for vascular risk and folate intervention studies.
Product Description
Supelco
Cyanocobalamin (B12), analytical standard
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Sigma-Aldrich
Cyanocobalamin, tested according to Ph. Eur.
Cyanocobalamin, European Pharmacopoeia (EP) Reference Standard
Supelco
Cyanocobalamin, pharmaceutical secondary standard, certified reference material
Cyanocobalamin, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%