Merck
  • Cobalt skin dose resulting from short and repetitive contact with hard metals.
Cobalt skin dose resulting from short and repetitive contact with hard metals.
Contact dermatitis (2014-03-13)
Klara Midander, Anneli Julander, Lizbet Skare, Carola Lidén
ABSTRACT
Many daily contacts with metallic items are short and repetitive, and result in metal release; material, sweat, friction and wear may all be important. To study cobalt release and skin deposition as a result of many short and repetitive contacts with two cobalt-containing materials. Study participants (n = 5) handled two types of hard metal disc (Co 6% and Co 15% Cr 0.6%) for 30 min. Deposited cobalt skin doses were measured with acid wipe sampling and chemical analysis. Cobalt release from the hard metal discs in artificial sweat was measured under conditions simulating those present during the handling of discs. Average cobalt skin doses from discs containing 6% Co and 15% Co 0.6% Cr were 1.1 µg/cm(2) [standard deviation (SD) 0.4 µg/cm(2) ] and 0.7 µg/cm(2) (SD 0.5 µg/cm(2) ), respectively. More cobalt was released from hard metal discs containing 6% Co [11.4 µg/cm(2) (SD 1.2 µg/cm(2) )] than from discs containing 15% Co 0.6% Cr [4.8 µg/cm(2) (SD 0.6 µg/cm(2) )]. 10% to 15% of the potentially available cobalt was deposited on the skin during handling. It is likely that the cobalt deposited as a result of short and repetitive contact with hard metals may cause harm. Research regarding cobalt exposure, metal release and the deposition of skin-sensitizing metals resulting from short and repetitive contact is needed for a better understanding of the allergy risk.
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Pricing and availability is not currently available.
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Pricing and availability is not currently available.
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Pricing and availability is not currently available.
Sigma-Aldrich
Nitric acid, puriss. p.a., ≥65% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Pricing and availability is not currently available.
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Pricing and availability is not currently available.