Merck
Clinical significance of CD155 expression in human pancreatic cancer.
Anticancer research (2015-04-12)
Satoshi Nishiwada, Masayuki Sho, Satoshi Yasuda, Keiji Shimada, Ichiro Yamato, Takahiro Akahori, Shoichi Kinoshita, Minako Nagai, Noboru Konishi, Yoshiyuki Nakajima
ABSTRACT
CD155 is expressed in many types of human cells and has diverse functions. We herein evaluated the clinical importance of CD155 in pancreatic cancer. We investigated CD155 expression in 134 patients with pancreatic cancer, and evaluated the correlations of CD155 with prognosis, tumor immunity and angiogenesis. Furthermore, CD155 functions were examined. CD155 expression was abundant in pancreatic cancer tissues. Patients with high CD155 expression had poorer postoperative prognosis than those with low expression. Multivariate analysis indicated that CD155 expression had a significant independent prognostic value. Tumor CD155 expression inversely correlated with the presence of tumor-infiltrating lymphocytes. Furthermore, it significantly positively correlated with vascular endothelial growth factor expression and intra tumoral microvessel density. In addition, silencing of CD155 inhibited proliferation, and induced cell-cycle arrest at G2/M phase in pancreatic cancer cells. CD155 may play a critical role through both immunological and non-immuno logical mechanisms in pancreatic cancer and may be a therapeutic target for this intractable malignancy.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Pvr
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
MISSION® esiRNA, targeting human PVR
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Adenosine 5′-diphosphoribose sodium salt, ≥93%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard