Merck
  • Use of the viability reagent PrestoBlue in comparison with alamarBlue and MTT to assess the viability of human corneal epithelial cells.
Use of the viability reagent PrestoBlue in comparison with alamarBlue and MTT to assess the viability of human corneal epithelial cells.
Journal of pharmacological and toxicological methods (2014-12-03)
Manlong Xu, David J McCanna, Jacob G Sivak
ABSTRACT
PrestoBlue is a new resazurin based reagent to assess cell viability and cytotoxicity. It is claimed to be a fast and highly sensitive assay. Here, we compared PrestoBlue, alamarBlue, and 3-[4,5-dimethylthiazol-2-yl]-2,5-diphenyl tetrazonium bromide (MTT) in assessing cell viability of human corneal epithelial cells (HCEC), and investigated the effect of plate color, reading mode, and plate storage on the performance of PrestoBlue assay. The viability of different numbers of healthy HCEC and the toxicity of various chemicals on HCEC were evaluated using PrestoBlue (fluorescence), alamarBlue (fluorescence), and MTT (absorbance). The sensitivities of the three assays were compared. In the PrestoBlue assay, three plate colors and two reading modes were used and compared in assessing the toxic effect of sodium dodecyl sulfate (SDS). The PrestoBlue solutions after reaction were stored and measured on day 1, 2, 3, 5, and 7. The fluorescence readings obtained on different days were then compared. Both PrestoBlue and alamarBlue were able to detect 5000 healthy cells after 30min incubation and 1000 cells after 1h, 2h, and 4h incubation; while MTT was able to detect 5000 cells after 3h incubation. In the assessment of the toxicity of various chemicals, PrestoBlue and alamarBlue performed similarly. There was no significant difference between the results obtained by these two reagents. All the three plate colors and two reading modes showed similar results in the PrestoBlue assay in assessing the toxicity of SDS. Plate storage up to 7days did not affect the result of the PrestoBlue assay. Our study suggests that in evaluating the viability of HCEC, PrestoBlue is more sensitive than MTT, but similar to alamarBlue. The plate color, reading mode and plate storage up to 7days did not affect the performance of the PrestoBlue assay.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard