Merck
  • Separation and purification of thymopentin with molecular imprinting membrane by solid phase extraction disks.
Separation and purification of thymopentin with molecular imprinting membrane by solid phase extraction disks.
Journal of pharmaceutical and biomedical analysis (2014-09-30)
Chaoli Wang, Xiaoling Hu, Ping Guan, Danfeng Wu, Liwei Qian, Ji Li, Renyuan Song
ABSTRACT
The synthesis and performance of molecularly imprinted membranes (MIMs) as a solid phase extraction packing materials for the separation and purification of thymopentin from crude samples was described. In order to increase structural selectivity and imprinting efficiency, surface-initiated ATRP and ionic liquid (1-vinyl-3-ethyl acetate imidazolium chloride) were used to prepare molecularly imprinting membranes. The results demonstrated that solid phase extraction disks stuffed by MIMs with ionic liquids as functional monomer demonstrated high isolation and purification of performance to the thymopentin. The molecular recognition of thymopentin was analyzed by using molecular modeling software.
Sigma-Aldrich
Copper(I) chloride, AnhydroBeads, ≥99.99% trace metals basis
Sigma-Aldrich
Copper(I) chloride, ≥99.995% trace metals basis
Sigma-Aldrich
Copper(I) chloride, reagent grade, 97%
Sigma-Aldrich
Copper(I) chloride, ReagentPlus®, purified, ≥99%
Sigma-Aldrich
α-Bromoisobutyryl bromide, 98%