Merck
  • A polysaccharide from Salvia miltiorrhiza Bunge improves immune function in gastric cancer rats.
A polysaccharide from Salvia miltiorrhiza Bunge improves immune function in gastric cancer rats.
Carbohydrate polymers (2014-07-20)
Nan Wang, Jingyue Yang, Jianguo Lu, Qing Qiao, Tao Wu, Xilin Du, Guoqiang Bao, Xianli He
ABSTRACT
A neutral polysaccharide fraction (SMPA) prepared from the roots of Salvia miltiorrhiza by DEAE-cellulose and Sephadex G-100 chromatography was tested for its immune enhancing function in N-methyl-N'-nitro-nitrosoguanidine (MNNG)-induced gastric cancer rats by intragastric administration. SMPA (200mg/kg) treatment not only increased the body weight, but also improved the immune organ indices. Furthermore, studies of various immunological activities in gastric cancer rats revealed that SMPA significantly stimulated splenocyte proliferation, promoted anti-inflammatory cytokines (IL-2, IL-4 and IL-10) production, inhibited pro-inflammatory cytokine (IL-6 and TNF-α) secretion, augmented the killing activity of natural killer (NK) cells and cytotoxic T lymphocytes (CTL), and increased phagocytotic function of macrophages in gastric cancer rats. In addition, SMPA administration evidently elevated total intracellular granzyme-B and IFN-γ levels produced by splenocytes in gastric cancer rats. Taken together, these results suggested that SMPA could act as an effective immunomodulator and might be explored as a potential supplemental source for gastric cancer therapy.
Product Description
Millipore
D-(−)-Arabinose, suitable for microbiology, ≥99.0%
Millipore
D-(+)-Galactose, suitable for microbiology, ≥99.0%
Millipore
D-(+)-Mannose, suitable for microbiology, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Millipore
D-(+)-Mannose, suitable for microbiology, ≥99%
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Sigma-Aldrich
D-(−)-Ribose, 98%
Sigma-Aldrich
D-(−)-Ribose, ≥98%
Sigma-Aldrich
D-Glucuronic acid, ≥98% (GC)
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(−)-Ribose, suitable for cell culture, BioReagent
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
D-(−)-Ribose, ≥99% (GC)
Sigma-Aldrich
D-(−)-Arabinose, ≥98% (GC)
Sigma-Aldrich
L-(−)-Fucose, ≥98% (GC)
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(+)-Mannose, ≥99% (GC), wood
Supelco
Galactose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99% (GC)
Sigma-Aldrich
Mouse IgM ELISA Kit
Xylose, European Pharmacopoeia (EP) Reference Standard
Galactose, European Pharmacopoeia (EP) Reference Standard
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
D-(+)-Xylose, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
D-(+)-Xylose, ≥99%