Merck
  • Glucose recognition proteins for glucose sensing at physiological concentrations and temperatures.
Glucose recognition proteins for glucose sensing at physiological concentrations and temperatures.
ACS chemical biology (2014-05-21)
Smita Joel, Kendrick B Turner, Sylvia Daunert
ABSTRACT
Advancements in biotechnology have allowed for the preparation of designer proteins with a wide spectrum of unprecedented chemical and physical properties. A variety of chemical and genetic methods can be employed to tailor the protein's properties, including its stability and various functions. Herein, we demonstrate the production of semisynthetic glucose recognition proteins (GRPs) prepared by truncating galactose/glucose binding protein (GBP) of E. coli and expanding the genetic code via global incorporation of unnatural amino acids into the structure of GBP and its fragments. The unnatural amino acids 5,5,5-trifluoroleucine (FL) and 5-fluorotryptophan (FW) were chosen for incorporation into the proteins. The resulting semisynthetic GRPs exhibit enhanced thermal stability and increased detection range of glucose without compromising its binding ability. These modifications enabled the utilization of the protein for the detection of glucose within physiological concentrations (mM) and temperatures ranging from hypothermia to hyperthermia. This ability to endow proteins such as GBP with improved stability and properties is critical in designing the next generation of tailor-made biosensing proteins for continuous in vivo glucose monitoring.
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
5-Fluoro-L-tryptophan, ≥98.0% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Supelco
Methanol, analytical standard
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium chloride solution, 0.85%