Merck
  • Aspergillus fumigatus triggers innate immune response via NOD1 signaling in human corneal epithelial cells.
Aspergillus fumigatus triggers innate immune response via NOD1 signaling in human corneal epithelial cells.
Experimental eye research (2014-08-12)
Yuting Zhang, Jiayin Wu, Zhaoting Xin, Xinyi Wu
ABSTRACT
Fungal keratitis is a serious vision-threatening disease caused by fungi after corneal epithelium damage. We have previously shown a role of cell surface TLRs in Aspergillus fumigatus (A. fumigatus) keratitis. In the present study we showed that Human telomerase-immortalized corneal epithelial cells (HCECs) exposed to A. fumigatus elicited an inflammatory response consisting in increased interleukin-6 (IL-6), IL-8 and tumor necrosis factor (TNF)-α expression and innate defense molecules hBD2 and LL37 in a time-dependent manner. In this study we further investigated the role of intracellular nucleotide-binding oligomerization domain-containing protein (NOD)-like receptors, NOD1 in innate immune and inflammatory response to A. fumigatus. We showed that NOD1 and its downstream signaling molecules RIP2 and NF-κB p65 are expressed in HCECs challenged with either NOD1 specific ligand iE-DAP or A. fumigatus. More importantly, NOD1 knockdown attenuated A. fumigatus-triggered the expression of NOD1, and downstream signaling effectors RIP2 and NF-κB p65, as well as the secretion of IL-6, IL-8 and TNF-α, and the production of hBD2 and LL37. In conclusion, our results demonstrated that NOD1 is a prominent factor of innate immune and inflammatory response in HCECs against A. fumigatus, suggesting that NOD1 might be a potential novel therapeutic target for the treatment of fungal keratitis.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Nod1
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture